Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 119895166-119895421 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAM30
Entrez Gene ID
11085
Gene Name
ADAM metallopeptidase domain 30
Gene Aliases
svph4
UniGene
Hs.283011
Gene Chromosome Location
Chr.1: 119893533-119896524 on Build GRCh38
Transcripts
NM_021794.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
894728 c.1164T>C p.C388C Chr.1: 119895173-119895173 on Build GRCh38
5526259 c.1068G>A p.R356R Chr.1: 119895269-119895269 on Build GRCh38
5508262 c.1059C>G p.C353W Chr.1: 119895278-119895278 on Build GRCh38
171176 c.1151C>T p.S384L Chr.1: 119895186-119895186 on Build GRCh38
458660 c.1058G>A p.C353Y Chr.1: 119895279-119895279 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774735574 Chr.1: 119895213-119895213 on Build GRCh38
rs781411347 Chr.1: 119895192-119895192 on Build GRCh38
rs587701025 Chr.1: 119895212-119895212 on Build GRCh38
rs41276632 Chr.1: 119895380-119895380 on Build GRCh38
rs777761187 Chr.1: 119895286-119895286 on Build GRCh38
rs202246434 Chr.1: 119895354-119895354 on Build GRCh38
rs769317336 Chr.1: 119895361-119895361 on Build GRCh38
rs773509669 Chr.1: 119895203-119895203 on Build GRCh38
rs775944924 Chr.1: 119895237-119895237 on Build GRCh38
rs766145128 Chr.1: 119895375-119895375 on Build GRCh38
rs936839708 Chr.1: 119895372-119895372 on Build GRCh38
rs139123456 Chr.1: 119895200-119895200 on Build GRCh38
rs369903502 Chr.1: 119895234-119895234 on Build GRCh38
rs374260098 Chr.1: 119895251-119895251 on Build GRCh38
rs752675578 Chr.1: 119895265-119895265 on Build GRCh38
rs762684242 Chr.1: 119895369-119895369 on Build GRCh38
rs377705019 Chr.1: 119895281-119895281 on Build GRCh38
rs993084456 Chr.1: 119895275-119895275 on Build GRCh38
rs992094249 Chr.1: 119895266-119895266 on Build GRCh38
rs945077516 Chr.1: 119895333-119895333 on Build GRCh38
rs759439950 Chr.1: 119895386-119895386 on Build GRCh38
rs772666710 Chr.1: 119895364-119895364 on Build GRCh38
rs749563971 Chr.1: 119895210-119895210 on Build GRCh38
rs749492038 Chr.1: 119895294-119895294 on Build GRCh38
rs2641348 Chr.1: 119895261-119895261 on Build GRCh38
rs148513488 Chr.1: 119895335-119895335 on Build GRCh38
rs767305753 Chr.1: 119895390-119895390 on Build GRCh38
rs760030228 Chr.1: 119895232-119895232 on Build GRCh38
rs754909170 Chr.1: 119895262-119895262 on Build GRCh38
rs776087621 Chr.1: 119895349-119895349 on Build GRCh38
rs768086429 Chr.1: 119895258-119895259 on Build GRCh38
rs962518414 Chr.1: 119895225-119895225 on Build GRCh38
rs759719895 Chr.1: 119895210-119895211 on Build GRCh38
rs941346359 Chr.1: 119895363-119895363 on Build GRCh38
rs761124638 Chr.1: 119895305-119895306 on Build GRCh38
rs750069068 Chr.1: 119895249-119895249 on Build GRCh38
rs774505840 Chr.1: 119895192-119895193 on Build GRCh38
rs746244883 Chr.1: 119895334-119895334 on Build GRCh38
rs781198225 Chr.1: 119895263-119895263 on Build GRCh38
rs761418593 Chr.1: 119895238-119895238 on Build GRCh38
rs748278094 Chr.1: 119895195-119895195 on Build GRCh38
rs917390514 Chr.1: 119895291-119895291 on Build GRCh38
rs1037007087 Chr.1: 119895388-119895388 on Build GRCh38
rs776040586 Chr.1: 119895271-119895271 on Build GRCh38
rs756236574 Chr.1: 119895275-119895275 on Build GRCh38
rs144550343 Chr.1: 119895328-119895328 on Build GRCh38
rs771208017 Chr.1: 119895303-119895303 on Build GRCh38
rs764747494 Chr.1: 119895242-119895242 on Build GRCh38
rs766274936 Chr.1: 119895253-119895253 on Build GRCh38

View Full Product Details