Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.Y: 6869993-6870248 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMELY
Entrez Gene ID
266
Gene Name
amelogenin, Y-linked
Gene Aliases
AMGL, AMGY
UniGene
Hs.1238
Gene Chromosome Location
Chr.Y: 6865918-6874056 on Build GRCh38
Transcripts
NM_001143.1 XM_017030042.1

SNP Detail

SNP ID Chromosome Location
rs945096224 Chr.Y: 6870110-6870110 on Build GRCh38
rs545301207 Chr.Y: 6870035-6870035 on Build GRCh38
rs375373860 Chr.Y: 6870102-6870102 on Build GRCh38

View Full Product Details