Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 44780986-44781243 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CBLC
Entrez Gene ID
23624
Gene Name
Cbl proto-oncogene C
Gene Aliases
CBL-3, CBL-SL, RNF57
UniGene
Hs.466907
Gene Chromosome Location
Chr.19: 44777855-44800646 on Build GRCh38
Transcripts
NM_001130852.1 NM_012116.3 XM_005258696.3 XM_011526688.2 XM_011526689.2 XM_011526690.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4517918 c.462_463CC>TT p.P155S Chr.19: 44781013-44781014 on Build GRCh38
6372983 c.482G>A p.R161K Chr.19: 44781033-44781033 on Build GRCh38
94016 c.497C>A p.A166D Chr.19: 44781048-44781048 on Build GRCh38
6043277 c.450G>A p.Q150Q Chr.19: 44781001-44781001 on Build GRCh38
6620921 c.493G>A p.G165R Chr.19: 44781044-44781044 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs111648890 Chr.19: 44781141-44781141 on Build GRCh38
rs752145860 Chr.19: 44781050-44781050 on Build GRCh38
rs774106147 Chr.19: 44781169-44781169 on Build GRCh38
rs753565633 Chr.19: 44781194-44781194 on Build GRCh38
rs750423011 Chr.19: 44781179-44781180 on Build GRCh38
rs757390586 Chr.19: 44781020-44781021 on Build GRCh38
rs41301961 Chr.19: 44781133-44781133 on Build GRCh38
rs772426257 Chr.19: 44781076-44781076 on Build GRCh38
rs200565443 Chr.19: 44781091-44781091 on Build GRCh38
rs1013092291 Chr.19: 44781103-44781103 on Build GRCh38
rs371124704 Chr.19: 44781024-44781024 on Build GRCh38
rs548291852 Chr.19: 44781104-44781104 on Build GRCh38
rs140442555 Chr.19: 44781044-44781044 on Build GRCh38
rs776720368 Chr.19: 44781101-44781101 on Build GRCh38
rs779369626 Chr.19: 44781066-44781066 on Build GRCh38
rs766214223 Chr.19: 44781164-44781164 on Build GRCh38
rs34511517 Chr.19: 44781109-44781110 on Build GRCh38
rs772149528 Chr.19: 44781192-44781192 on Build GRCh38
rs751522770 Chr.19: 44781043-44781043 on Build GRCh38
rs971205358 Chr.19: 44781113-44781113 on Build GRCh38
rs570439935 Chr.19: 44781180-44781180 on Build GRCh38
rs917964274 Chr.19: 44781157-44781157 on Build GRCh38
rs746425994 Chr.19: 44781067-44781067 on Build GRCh38
rs755586372 Chr.19: 44781189-44781189 on Build GRCh38
rs775409459 Chr.19: 44781011-44781011 on Build GRCh38
rs372353828 Chr.19: 44781176-44781176 on Build GRCh38
rs758998572 Chr.19: 44781216-44781216 on Build GRCh38
rs762748099 Chr.19: 44781027-44781027 on Build GRCh38
rs553276640 Chr.19: 44781219-44781219 on Build GRCh38
rs748884728 Chr.19: 44781054-44781054 on Build GRCh38
rs35106910 Chr.19: 44781009-44781009 on Build GRCh38
rs201056899 Chr.19: 44781078-44781078 on Build GRCh38
rs1027715206 Chr.19: 44781074-44781074 on Build GRCh38
rs761978098 Chr.19: 44781163-44781163 on Build GRCh38
rs909102893 Chr.19: 44781177-44781177 on Build GRCh38
rs182379358 Chr.19: 44781092-44781092 on Build GRCh38
rs780644821 Chr.19: 44781211-44781212 on Build GRCh38
rs746229759 Chr.19: 44781209-44781209 on Build GRCh38
rs375598616 Chr.19: 44781178-44781178 on Build GRCh38
rs779460411 Chr.19: 44781206-44781206 on Build GRCh38
rs114493128 Chr.19: 44781062-44781062 on Build GRCh38
rs201807125 Chr.19: 44781051-44781051 on Build GRCh38
rs201551471 Chr.19: 44781016-44781016 on Build GRCh38
rs202052679 Chr.19: 44781200-44781200 on Build GRCh38
rs530653216 Chr.19: 44781140-44781140 on Build GRCh38
rs756676757 Chr.19: 44781183-44781193 on Build GRCh38
rs764214326 Chr.19: 44781017-44781017 on Build GRCh38
rs747042705 Chr.19: 44781079-44781079 on Build GRCh38
rs767098851 Chr.19: 44781046-44781046 on Build GRCh38
rs767685481 Chr.19: 44781075-44781076 on Build GRCh38
rs186238294 Chr.19: 44781093-44781093 on Build GRCh38
rs766093151 Chr.19: 44781040-44781040 on Build GRCh38
rs993192049 Chr.19: 44781070-44781070 on Build GRCh38

View Full Product Details