Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 10291075-10291344 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ICAM5
Entrez Gene ID
7087
Gene Name
intercellular adhesion molecule 5
Gene Aliases
TLCN, TLN
UniGene
Hs.465862
Gene Chromosome Location
Chr.19: 10289352-10296778 on Build GRCh38
Transcripts
NM_003259.3 XM_011528229.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7157873 c.181C>T p.R61W Chr.19: 10291170-10291170 on Build GRCh38
6300494 c.293T>A p.V98D Chr.19: 10291282-10291282 on Build GRCh38
4073783 c.128C>T p.A43V Chr.19: 10291117-10291117 on Build GRCh38
1241288 c.122G>A p.R41H Chr.19: 10291111-10291111 on Build GRCh38
710251 c.187G>C p.E63Q Chr.19: 10291176-10291176 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753009545 Chr.19: 10291238-10291238 on Build GRCh38
rs745979453 Chr.19: 10291281-10291281 on Build GRCh38
rs56406350 Chr.19: 10291220-10291220 on Build GRCh38
rs11669397 Chr.19: 10291170-10291170 on Build GRCh38
rs775835159 Chr.19: 10291292-10291292 on Build GRCh38
rs756477605 Chr.19: 10291241-10291241 on Build GRCh38
rs748812071 Chr.19: 10291236-10291236 on Build GRCh38
rs779073267 Chr.19: 10291262-10291262 on Build GRCh38
rs1049631085 Chr.19: 10291111-10291111 on Build GRCh38
rs768495461 Chr.19: 10291298-10291298 on Build GRCh38
rs764790873 Chr.19: 10291182-10291182 on Build GRCh38
rs761753686 Chr.19: 10291308-10291308 on Build GRCh38
rs372844196 Chr.19: 10291256-10291256 on Build GRCh38
rs779711819 Chr.19: 10291232-10291232 on Build GRCh38
rs887845645 Chr.19: 10291279-10291279 on Build GRCh38
rs757978671 Chr.19: 10291199-10291199 on Build GRCh38
rs377323998 Chr.19: 10291293-10291293 on Build GRCh38
rs763846097 Chr.19: 10291100-10291100 on Build GRCh38
rs749793668 Chr.19: 10291260-10291260 on Build GRCh38
rs760572744 Chr.19: 10291096-10291096 on Build GRCh38
rs765091105 Chr.19: 10291310-10291310 on Build GRCh38
rs369518200 Chr.19: 10291142-10291142 on Build GRCh38
rs149309075 Chr.19: 10291261-10291261 on Build GRCh38
rs749948901 Chr.19: 10291195-10291195 on Build GRCh38
rs778036058 Chr.19: 10291244-10291244 on Build GRCh38
rs753779713 Chr.19: 10291161-10291161 on Build GRCh38
rs776348928 Chr.19: 10291306-10291306 on Build GRCh38
rs761723946 Chr.19: 10291163-10291163 on Build GRCh38
rs749895372 Chr.19: 10291318-10291318 on Build GRCh38
rs772374571 Chr.19: 10291282-10291282 on Build GRCh38

View Full Product Details