Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 155139801-155140060 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPM3
Entrez Gene ID
54344
Gene Name
dolichyl-phosphate mannosyltransferase subunit 3
Gene Aliases
CDG1O
UniGene
Hs.110477
Gene Chromosome Location
Chr.1: 155139891-155140573 on Build GRCh38
Transcripts
NM_018973.3 NM_153741.1 XM_017001498.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2209932 c.327G>A p.Q109Q Chr.1: 155140004-155140004 on Build GRCh38
3474859 c.331G>T p.A111S Chr.1: 155140000-155140000 on Build GRCh38
4847738 c.366C>T p.F122F Chr.1: 155139965-155139965 on Build GRCh38
1319917 c.315G>A p.Q105Q Chr.1: 155140016-155140016 on Build GRCh38
4022937 c.363C>T p.R121R Chr.1: 155139968-155139968 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749000879 Chr.1: 155139997-155139997 on Build GRCh38
rs758821383 Chr.1: 155139975-155139975 on Build GRCh38
rs1033351199 Chr.1: 155139854-155139854 on Build GRCh38
rs770729851 Chr.1: 155139933-155139933 on Build GRCh38
rs568001373 Chr.1: 155139964-155139964 on Build GRCh38
rs145285403 Chr.1: 155140009-155140009 on Build GRCh38
rs761931137 Chr.1: 155140034-155140034 on Build GRCh38
rs776752576 Chr.1: 155139960-155139960 on Build GRCh38
rs777586354 Chr.1: 155139993-155139993 on Build GRCh38
rs912122244 Chr.1: 155139902-155139902 on Build GRCh38
rs929518101 Chr.1: 155139886-155139886 on Build GRCh38
rs200477845 Chr.1: 155139952-155139952 on Build GRCh38
rs750763940 Chr.1: 155139973-155139973 on Build GRCh38
rs752119871 Chr.1: 155139984-155139984 on Build GRCh38
rs760608219 Chr.1: 155139945-155139945 on Build GRCh38
rs114265504 Chr.1: 155139862-155139862 on Build GRCh38
rs747698691 Chr.1: 155139991-155139991 on Build GRCh38
rs765483911 Chr.1: 155139970-155139970 on Build GRCh38
rs745807155 Chr.1: 155140023-155140023 on Build GRCh38
rs959069799 Chr.1: 155139857-155139857 on Build GRCh38
rs193215070 Chr.1: 155139992-155139992 on Build GRCh38
rs755494157 Chr.1: 155139985-155139985 on Build GRCh38
rs987542488 Chr.1: 155139890-155139890 on Build GRCh38
rs770767631 Chr.1: 155140006-155140006 on Build GRCh38
rs752974130 Chr.1: 155139846-155139846 on Build GRCh38
rs773107257 Chr.1: 155139922-155139922 on Build GRCh38
rs981747581 Chr.1: 155139845-155139845 on Build GRCh38
rs766728407 Chr.1: 155139977-155139977 on Build GRCh38
rs774380780 Chr.1: 155139943-155139943 on Build GRCh38
rs772066343 Chr.1: 155140029-155140029 on Build GRCh38
rs121908155 Chr.1: 155139987-155139987 on Build GRCh38
rs528199014 Chr.1: 155139930-155139930 on Build GRCh38
rs920751139 Chr.1: 155139871-155139871 on Build GRCh38
rs913390061 Chr.1: 155139904-155139904 on Build GRCh38
rs776662775 Chr.1: 155140033-155140033 on Build GRCh38
rs138800426 Chr.1: 155139888-155139888 on Build GRCh38
rs535242424 Chr.1: 155140039-155140039 on Build GRCh38
rs762019309 Chr.1: 155139961-155139961 on Build GRCh38

View Full Product Details