Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 30784136-30784409 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRLF3
Entrez Gene ID
51379
Gene Name
cytokine receptor like factor 3
Gene Aliases
CREME-9, CREME9, CRLM9, CYTOR4, FRWS, p48.2
UniGene
Hs.649372
Gene Chromosome Location
Chr.17: 30782684-30824760 on Build GRCh38
Transcripts
NM_015986.3 NR_073118.1 XM_011524879.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6363679 c.1154A>G p.E385G Chr.17: 30784362-30784362 on Build GRCh38
5047800 c.1216A>G p.I406V Chr.17: 30784300-30784300 on Build GRCh38
705705 c.1309T>C p.W437R Chr.17: 30784207-30784207 on Build GRCh38
1381969 c.1234G>A p.E412K Chr.17: 30784282-30784282 on Build GRCh38
5129505 c.1158C>T p.A386A Chr.17: 30784358-30784358 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs554716980 Chr.17: 30784213-30784213 on Build GRCh38
rs748397239 Chr.17: 30784373-30784373 on Build GRCh38
rs778909575 Chr.17: 30784268-30784268 on Build GRCh38
rs751235967 Chr.17: 30784341-30784341 on Build GRCh38
rs34688098 Chr.17: 30784339-30784340 on Build GRCh38
rs377421958 Chr.17: 30784229-30784229 on Build GRCh38
rs766976106 Chr.17: 30784387-30784387 on Build GRCh38
rs563742674 Chr.17: 30784358-30784358 on Build GRCh38
rs754084666 Chr.17: 30784211-30784211 on Build GRCh38
rs201413764 Chr.17: 30784169-30784169 on Build GRCh38
rs761170871 Chr.17: 30784324-30784324 on Build GRCh38
rs200481377 Chr.17: 30784174-30784174 on Build GRCh38
rs11867457 Chr.17: 30784350-30784350 on Build GRCh38
rs745823775 Chr.17: 30784250-30784250 on Build GRCh38
rs774434726 Chr.17: 30784300-30784300 on Build GRCh38
rs201887817 Chr.17: 30784338-30784338 on Build GRCh38
rs987945475 Chr.17: 30784382-30784382 on Build GRCh38
rs202178927 Chr.17: 30784165-30784165 on Build GRCh38
rs749176167 Chr.17: 30784355-30784355 on Build GRCh38
rs769690770 Chr.17: 30784157-30784157 on Build GRCh38
rs770536701 Chr.17: 30784244-30784244 on Build GRCh38
rs374412323 Chr.17: 30784202-30784202 on Build GRCh38
rs751676888 Chr.17: 30784181-30784181 on Build GRCh38
rs775766885 Chr.17: 30784327-30784329 on Build GRCh38
rs754286419 Chr.17: 30784335-30784335 on Build GRCh38
rs780423759 Chr.17: 30784201-30784201 on Build GRCh38
rs899852304 Chr.17: 30784255-30784255 on Build GRCh38
rs371052725 Chr.17: 30784259-30784259 on Build GRCh38
rs750904788 Chr.17: 30784198-30784198 on Build GRCh38
rs77538147 Chr.17: 30784363-30784363 on Build GRCh38
rs572945703 Chr.17: 30784226-30784226 on Build GRCh38
rs34159788 Chr.17: 30784316-30784317 on Build GRCh38
rs36124437 Chr.17: 30784180-30784181 on Build GRCh38
rs113946501 Chr.17: 30784186-30784186 on Build GRCh38
rs767602957 Chr.17: 30784306-30784306 on Build GRCh38
rs781329571 Chr.17: 30784248-30784248 on Build GRCh38
rs766738527 Chr.17: 30784334-30784334 on Build GRCh38
rs1030131472 Chr.17: 30784331-30784331 on Build GRCh38
rs139807989 Chr.17: 30784357-30784357 on Build GRCh38
rs199632265 Chr.17: 30784175-30784175 on Build GRCh38
rs780880303 Chr.17: 30784347-30784347 on Build GRCh38
rs1039682387 Chr.17: 30784249-30784249 on Build GRCh38
rs763849169 Chr.17: 30784340-30784340 on Build GRCh38
rs779846248 Chr.17: 30784353-30784353 on Build GRCh38
rs773292810 Chr.17: 30784374-30784374 on Build GRCh38
rs756137945 Chr.17: 30784352-30784352 on Build GRCh38
rs200013016 Chr.17: 30784288-30784288 on Build GRCh38
rs756560673 Chr.17: 30784199-30784199 on Build GRCh38
rs770603773 Chr.17: 30784300-30784300 on Build GRCh38
rs756922692 Chr.17: 30784343-30784343 on Build GRCh38
rs773231690 Chr.17: 30784313-30784313 on Build GRCh38
rs768050686 Chr.17: 30784187-30784187 on Build GRCh38
rs760729327 Chr.17: 30784378-30784378 on Build GRCh38
rs764159202 Chr.17: 30784172-30784172 on Build GRCh38
rs775099954 Chr.17: 30784158-30784158 on Build GRCh38
rs111760885 Chr.17: 30784258-30784258 on Build GRCh38

View Full Product Details