Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 56452523-56452770 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASS4
Entrez Gene ID
57091
Gene Name
Cas scaffolding protein family member 4
Gene Aliases
C20orf32, CAS4, HEFL, HEPL
UniGene
Hs.473144
Gene Chromosome Location
Chr.20: 56411550-56460240 on Build GRCh38
Transcripts
NM_001164114.1 NM_001164115.1 NM_001164116.1 NM_020356.3 XM_006723831.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2763995 c.1542C>G p.N514K Chr.20: 56452718-56452718 on Build GRCh38
3548044 c.1500C>T p.V500V Chr.20: 56452676-56452676 on Build GRCh38
1412630 c.1363G>A p.V455I Chr.20: 56452539-56452539 on Build GRCh38
1028281 c.1444T>C p.S482P Chr.20: 56452620-56452620 on Build GRCh38
1412631 c.1493G>A p.R498Q Chr.20: 56452669-56452669 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771080322 Chr.20: 56452668-56452668 on Build GRCh38
rs144538897 Chr.20: 56452562-56452562 on Build GRCh38
rs919291093 Chr.20: 56452565-56452565 on Build GRCh38
rs753526844 Chr.20: 56452582-56452582 on Build GRCh38
rs1044586660 Chr.20: 56452660-56452660 on Build GRCh38
rs891477283 Chr.20: 56452726-56452726 on Build GRCh38
rs747621928 Chr.20: 56452619-56452619 on Build GRCh38
rs761184541 Chr.20: 56452742-56452742 on Build GRCh38
rs749307548 Chr.20: 56452665-56452665 on Build GRCh38
rs762367653 Chr.20: 56452729-56452729 on Build GRCh38
rs764139797 Chr.20: 56452722-56452722 on Build GRCh38
rs760752577 Chr.20: 56452717-56452717 on Build GRCh38
rs755326072 Chr.20: 56452641-56452641 on Build GRCh38
rs201454304 Chr.20: 56452659-56452659 on Build GRCh38
rs775284457 Chr.20: 56452669-56452669 on Build GRCh38
rs756912515 Chr.20: 56452586-56452586 on Build GRCh38
rs749857293 Chr.20: 56452592-56452592 on Build GRCh38
rs550251334 Chr.20: 56452735-56452735 on Build GRCh38
rs917517655 Chr.20: 56452608-56452608 on Build GRCh38
rs564505566 Chr.20: 56452724-56452724 on Build GRCh38
rs764045834 Chr.20: 56452568-56452568 on Build GRCh38
rs371150425 Chr.20: 56452728-56452728 on Build GRCh38
rs764981924 Chr.20: 56452591-56452591 on Build GRCh38
rs533336238 Chr.20: 56452725-56452725 on Build GRCh38
rs904655774 Chr.20: 56452696-56452696 on Build GRCh38
rs1044229460 Chr.20: 56452587-56452587 on Build GRCh38
rs757945426 Chr.20: 56452601-56452601 on Build GRCh38
rs146122261 Chr.20: 56452603-56452603 on Build GRCh38
rs770021620 Chr.20: 56452559-56452559 on Build GRCh38
rs16979936 Chr.20: 56452648-56452648 on Build GRCh38
rs527807357 Chr.20: 56452548-56452548 on Build GRCh38
rs145762733 Chr.20: 56452651-56452651 on Build GRCh38
rs1029721066 Chr.20: 56452751-56452751 on Build GRCh38
rs140151545 Chr.20: 56452622-56452622 on Build GRCh38
rs866071361 Chr.20: 56452650-56452650 on Build GRCh38
rs776832429 Chr.20: 56452543-56452543 on Build GRCh38
rs376814976 Chr.20: 56452566-56452566 on Build GRCh38
rs770317129 Chr.20: 56452644-56452644 on Build GRCh38
rs559542055 Chr.20: 56452542-56452542 on Build GRCh38
rs377729881 Chr.20: 56452740-56452740 on Build GRCh38
rs767310064 Chr.20: 56452564-56452564 on Build GRCh38
rs138464146 Chr.20: 56452694-56452694 on Build GRCh38

View Full Product Details