Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 53864862-53865123 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTR8
Entrez Gene ID
93973
Gene Name
ARP8 actin-related protein 8 homolog
Gene Aliases
ARP8, INO80N, hArp8
UniGene
Hs.412186
Gene Chromosome Location
Chr.3: 53863096-53882202 on Build GRCh38
Transcripts
XM_005265587.4 NM_018725.3 XM_005265310.4 XM_005265311.4 XM_005265312.4 XM_011533941.2 XM_017006804.1 XM_017006805.1 XM_017006806.1 XM_017006807.1

Gene Symbol
IL17RB
Entrez Gene ID
55540
Gene Name
interleukin 17 receptor B
Gene Aliases
CRL4, EVI27, IL17BR, IL17RH1
UniGene
Hs.654970
Gene Chromosome Location
Chr.3: 53846494-53866526 on Build GRCh38
Transcripts
XM_005265587.4 NM_018725.3 XM_005265310.4 XM_005265311.4 XM_005265312.4 XM_011533941.2 XM_017006804.1 XM_017006805.1 XM_017006806.1 XM_017006807.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2781043 c.1088T>A p.V363D Chr.3: 53864887-53864887 on Build GRCh38
1047334 c.1261G>T p.D421Y Chr.3: 53865060-53865060 on Build GRCh38
4841707 c.1290C>G p.F430L Chr.3: 53865089-53865089 on Build GRCh38
5720052 c.1302A>C p.L434L Chr.3: 53865101-53865101 on Build GRCh38
3427768 c.1172A>C p.K391T Chr.3: 53864971-53864971 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747392178 Chr.3: 53865068-53865068 on Build GRCh38
rs768551820 Chr.3: 53865084-53865085 on Build GRCh38
rs201219894 Chr.3: 53864957-53864957 on Build GRCh38
rs770278772 Chr.3: 53864965-53864968 on Build GRCh38
rs954661861 Chr.3: 53865052-53865052 on Build GRCh38
rs372602299 Chr.3: 53865025-53865025 on Build GRCh38
rs763699823 Chr.3: 53865029-53865029 on Build GRCh38
rs759547152 Chr.3: 53864906-53864906 on Build GRCh38
rs895231805 Chr.3: 53864949-53864949 on Build GRCh38
rs769401500 Chr.3: 53865015-53865015 on Build GRCh38
rs555347777 Chr.3: 53865094-53865094 on Build GRCh38
rs143904356 Chr.3: 53865030-53865030 on Build GRCh38
rs761863476 Chr.3: 53865032-53865032 on Build GRCh38
rs199637339 Chr.3: 53864997-53864997 on Build GRCh38
rs750609380 Chr.3: 53864992-53864992 on Build GRCh38
rs987407603 Chr.3: 53865056-53865056 on Build GRCh38
rs370331617 Chr.3: 53865073-53865073 on Build GRCh38
rs3733078 Chr.3: 53865011-53865011 on Build GRCh38
rs753001735 Chr.3: 53864969-53864969 on Build GRCh38
rs577437282 Chr.3: 53864975-53864975 on Build GRCh38
rs759137828 Chr.3: 53864917-53864917 on Build GRCh38
rs756335321 Chr.3: 53864993-53864993 on Build GRCh38
rs200317910 Chr.3: 53864946-53864946 on Build GRCh38
rs779327768 Chr.3: 53865008-53865008 on Build GRCh38
rs758868919 Chr.3: 53865058-53865058 on Build GRCh38
rs745502642 Chr.3: 53865012-53865012 on Build GRCh38
rs764783124 Chr.3: 53864944-53864944 on Build GRCh38
rs753185213 Chr.3: 53865050-53865050 on Build GRCh38
rs755066477 Chr.3: 53865007-53865007 on Build GRCh38
rs199790259 Chr.3: 53865048-53865048 on Build GRCh38
rs376992001 Chr.3: 53865065-53865065 on Build GRCh38
rs1006643031 Chr.3: 53864919-53864919 on Build GRCh38
rs780149135 Chr.3: 53864996-53864996 on Build GRCh38
rs200188637 Chr.3: 53865028-53865028 on Build GRCh38
rs746986147 Chr.3: 53864955-53864957 on Build GRCh38
rs766565095 Chr.3: 53865047-53865047 on Build GRCh38
rs748689565 Chr.3: 53865010-53865010 on Build GRCh38
rs749453165 Chr.3: 53864976-53864976 on Build GRCh38
rs760943596 Chr.3: 53865043-53865043 on Build GRCh38
rs377005084 Chr.3: 53864926-53864926 on Build GRCh38
rs776312127 Chr.3: 53864903-53864903 on Build GRCh38
rs2232349 Chr.3: 53864951-53864951 on Build GRCh38
rs767246780 Chr.3: 53864983-53864983 on Build GRCh38
rs978342154 Chr.3: 53864970-53864970 on Build GRCh38
rs774474582 Chr.3: 53864893-53864893 on Build GRCh38
rs188917846 Chr.3: 53865042-53865042 on Build GRCh38
rs767317623 Chr.3: 53865033-53865033 on Build GRCh38
rs374050382 Chr.3: 53865082-53865082 on Build GRCh38

View Full Product Details