Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 66633062-66633313 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCTD7
Entrez Gene ID
154881
Gene Name
potassium channel tetramerization domain containing 7
Gene Aliases
CLN14, EPM3
UniGene
Hs.546627
Gene Chromosome Location
Chr.7: 66628881-66643229 on Build GRCh38
Transcripts
NM_001167961.2 NM_153033.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1091327 c.162C>T p.P54P Chr.7: 66633292-66633292 on Build GRCh38
6178377 c.172G>T p.G58* Chr.7: 66633302-66633302 on Build GRCh38
6883532 c.160C>T p.P54S Chr.7: 66633290-66633290 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs375139826 Chr.7: 66633127-66633127 on Build GRCh38
rs150889525 Chr.7: 66633098-66633098 on Build GRCh38
rs368755155 Chr.7: 66633271-66633271 on Build GRCh38
rs770973872 Chr.7: 66633270-66633270 on Build GRCh38
rs776037212 Chr.7: 66633290-66633290 on Build GRCh38
rs75774733 Chr.7: 66633155-66633155 on Build GRCh38
rs781014688 Chr.7: 66633242-66633242 on Build GRCh38
rs1029536208 Chr.7: 66633089-66633089 on Build GRCh38
rs543103689 Chr.7: 66633256-66633256 on Build GRCh38
rs563740472 Chr.7: 66633191-66633191 on Build GRCh38
rs768463613 Chr.7: 66633241-66633241 on Build GRCh38
rs779862565 Chr.7: 66633238-66633238 on Build GRCh38
rs990584993 Chr.7: 66633194-66633194 on Build GRCh38
rs146147853 Chr.7: 66633174-66633174 on Build GRCh38
rs930526100 Chr.7: 66633259-66633259 on Build GRCh38
rs559020294 Chr.7: 66633275-66633275 on Build GRCh38
rs374702986 Chr.7: 66633254-66633254 on Build GRCh38
rs749343374 Chr.7: 66633260-66633260 on Build GRCh38
rs761117000 Chr.7: 66633292-66633292 on Build GRCh38
rs747910548 Chr.7: 66633248-66633248 on Build GRCh38
rs984736733 Chr.7: 66633105-66633105 on Build GRCh38
rs768112652 Chr.7: 66633280-66633280 on Build GRCh38
rs75054368 Chr.7: 66633154-66633154 on Build GRCh38
rs1012235730 Chr.7: 66633140-66633140 on Build GRCh38
rs3764905 Chr.7: 66633239-66633239 on Build GRCh38

View Full Product Details