Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 159806501-159806736 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FCRL6
Entrez Gene ID
343413
Gene Name
Fc receptor like 6
Gene Aliases
FcRH6
UniGene
Hs.196955
Gene Chromosome Location
Chr.1: 159800506-159816257 on Build GRCh38
Transcripts
NM_001004310.2 NM_001284217.1 XM_005245128.3 XM_005245129.3 XM_005245131.3 XM_006711292.3 XM_011509480.1 XM_011509481.2 XM_017001176.1 XM_017001177.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5477015 c.32-9C>A p.? Chr.1: 159806587-159806587 on Build GRCh38
5157274 c.32-8T>A p.? Chr.1: 159806588-159806588 on Build GRCh38
2084703 c.40G>A p.V14I Chr.1: 159806604-159806604 on Build GRCh38
2084702 c.34C>T p.P12S Chr.1: 159806598-159806598 on Build GRCh38
5594587 c.44G>A p.G15E Chr.1: 159806608-159806608 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs946878479 Chr.1: 159806663-159806663 on Build GRCh38
rs777732801 Chr.1: 159806621-159806621 on Build GRCh38
rs995097317 Chr.1: 159806552-159806552 on Build GRCh38
rs777144459 Chr.1: 159806567-159806567 on Build GRCh38
rs992864730 Chr.1: 159806706-159806706 on Build GRCh38
rs777087632 Chr.1: 159806668-159806668 on Build GRCh38
rs888187954 Chr.1: 159806529-159806529 on Build GRCh38
rs770464848 Chr.1: 159806652-159806652 on Build GRCh38
rs1029971650 Chr.1: 159806587-159806587 on Build GRCh38
rs750942960 Chr.1: 159806573-159806573 on Build GRCh38
rs377578549 Chr.1: 159806533-159806533 on Build GRCh38
rs760602710 Chr.1: 159806678-159806678 on Build GRCh38
rs964598614 Chr.1: 159806534-159806534 on Build GRCh38
rs144415344 Chr.1: 159806572-159806572 on Build GRCh38
rs576488123 Chr.1: 159806702-159806702 on Build GRCh38
rs958673587 Chr.1: 159806688-159806688 on Build GRCh38
rs745450633 Chr.1: 159806661-159806661 on Build GRCh38
rs766425671 Chr.1: 159806580-159806580 on Build GRCh38
rs747955875 Chr.1: 159806615-159806615 on Build GRCh38
rs761899314 Chr.1: 159806652-159806652 on Build GRCh38
rs980018387 Chr.1: 159806677-159806677 on Build GRCh38
rs772715372 Chr.1: 159806608-159806608 on Build GRCh38
rs1052762269 Chr.1: 159806712-159806712 on Build GRCh38
rs1005347757 Chr.1: 159806530-159806530 on Build GRCh38
rs751746440 Chr.1: 159806593-159806593 on Build GRCh38
rs1035019416 Chr.1: 159806624-159806624 on Build GRCh38
rs868708191 Chr.1: 159806595-159806595 on Build GRCh38
rs368159195 Chr.1: 159806637-159806637 on Build GRCh38
rs112211786 Chr.1: 159806655-159806655 on Build GRCh38
rs978780047 Chr.1: 159806685-159806685 on Build GRCh38
rs375691064 Chr.1: 159806662-159806662 on Build GRCh38
rs184922930 Chr.1: 159806710-159806710 on Build GRCh38
rs192211995 Chr.1: 159806683-159806683 on Build GRCh38
rs61821626 Chr.1: 159806576-159806576 on Build GRCh38
rs764499599 Chr.1: 159806561-159806561 on Build GRCh38
rs762370352 Chr.1: 159806571-159806571 on Build GRCh38
rs927394402 Chr.1: 159806687-159806687 on Build GRCh38
rs572690132 Chr.1: 159806619-159806619 on Build GRCh38
rs899467164 Chr.1: 159806568-159806568 on Build GRCh38
rs368413465 Chr.1: 159806539-159806539 on Build GRCh38
rs759323376 Chr.1: 159806558-159806558 on Build GRCh38
rs755197300 Chr.1: 159806601-159806601 on Build GRCh38

View Full Product Details