Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 102945659-102945928 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MMP13
Entrez Gene ID
4322
Gene Name
matrix metallopeptidase 13
Gene Aliases
CLG3, MANDP1, MMP-13
UniGene
Hs.2936
Gene Chromosome Location
Chr.11: 102942992-102955734 on Build GRCh38
Transcripts
NM_002427.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2108567 c.1231A>T p.I411F Chr.11: 102945730-102945730 on Build GRCh38
4524777 c.1291G>A p.V431I Chr.11: 102945670-102945670 on Build GRCh38
922312 c.1251G>A p.P417P Chr.11: 102945710-102945710 on Build GRCh38
4524351 c.1267G>A p.D423N Chr.11: 102945694-102945694 on Build GRCh38
1350427 c.1225A>G p.N409D Chr.11: 102945736-102945736 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782188014 Chr.11: 102945800-102945800 on Build GRCh38
rs781834016 Chr.11: 102945775-102945775 on Build GRCh38
rs912940258 Chr.11: 102945877-102945877 on Build GRCh38
rs957593499 Chr.11: 102945858-102945858 on Build GRCh38
rs572644245 Chr.11: 102945724-102945724 on Build GRCh38
rs782792053 Chr.11: 102945814-102945814 on Build GRCh38
rs544466500 Chr.11: 102945857-102945857 on Build GRCh38
rs782277300 Chr.11: 102945760-102945760 on Build GRCh38
rs1014231242 Chr.11: 102945811-102945811 on Build GRCh38
rs781897909 Chr.11: 102945878-102945878 on Build GRCh38
rs553095955 Chr.11: 102945868-102945868 on Build GRCh38
rs1040017514 Chr.11: 102945712-102945712 on Build GRCh38
rs781885988 Chr.11: 102945768-102945768 on Build GRCh38
rs782686092 Chr.11: 102945770-102945770 on Build GRCh38
rs782457233 Chr.11: 102945782-102945782 on Build GRCh38
rs782543033 Chr.11: 102945794-102945794 on Build GRCh38
rs980373775 Chr.11: 102945822-102945822 on Build GRCh38
rs781872133 Chr.11: 102945762-102945762 on Build GRCh38
rs782044398 Chr.11: 102945733-102945733 on Build GRCh38
rs782592871 Chr.11: 102945797-102945797 on Build GRCh38
rs368922836 Chr.11: 102945750-102945750 on Build GRCh38
rs782604201 Chr.11: 102945721-102945721 on Build GRCh38
rs150203792 Chr.11: 102945710-102945710 on Build GRCh38
rs988963337 Chr.11: 102945875-102945875 on Build GRCh38
rs782542049 Chr.11: 102945689-102945689 on Build GRCh38
rs782680283 Chr.11: 102945759-102945760 on Build GRCh38
rs782645077 Chr.11: 102945783-102945783 on Build GRCh38
rs190896822 Chr.11: 102945747-102945747 on Build GRCh38
rs782304020 Chr.11: 102945784-102945784 on Build GRCh38
rs782157940 Chr.11: 102945740-102945740 on Build GRCh38
rs961763199 Chr.11: 102945798-102945798 on Build GRCh38
rs554838961 Chr.11: 102945763-102945763 on Build GRCh38
rs564571817 Chr.11: 102945769-102945769 on Build GRCh38
rs182374215 Chr.11: 102945828-102945828 on Build GRCh38
rs782099092 Chr.11: 102945749-102945749 on Build GRCh38
rs970274853 Chr.11: 102945818-102945818 on Build GRCh38
rs782812377 Chr.11: 102945767-102945767 on Build GRCh38
rs36086947 Chr.11: 102945686-102945687 on Build GRCh38

View Full Product Details