Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 101031709-101031980 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MUC17
Entrez Gene ID
140453
Gene Name
mucin 17, cell surface associated
Gene Aliases
MUC-17, MUC-3, MUC3
UniGene
Hs.271819
Gene Chromosome Location
Chr.7: 101020083-101058859 on Build GRCh38
Transcripts
NM_001040105.1 NR_133665.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7189595 c.299C>T p.S100L Chr.7: 101031715-101031715 on Build GRCh38
5543152 c.510C>T p.P170P Chr.7: 101031926-101031926 on Build GRCh38
7272798 c.458G>A p.S153N Chr.7: 101031874-101031874 on Build GRCh38
7419731 c.319A>T p.S107C Chr.7: 101031735-101031735 on Build GRCh38
1158815 c.420A>T p.T140T Chr.7: 101031836-101031836 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772827847 Chr.7: 101031915-101031915 on Build GRCh38
rs761222920 Chr.7: 101031904-101031904 on Build GRCh38
rs372923916 Chr.7: 101031946-101031946 on Build GRCh38
rs745658408 Chr.7: 101031796-101031796 on Build GRCh38
rs372743652 Chr.7: 101031758-101031758 on Build GRCh38
rs375764950 Chr.7: 101031819-101031819 on Build GRCh38
rs775546915 Chr.7: 101031803-101031803 on Build GRCh38
rs763622974 Chr.7: 101031814-101031814 on Build GRCh38
rs766732211 Chr.7: 101031833-101031833 on Build GRCh38
rs201566918 Chr.7: 101031783-101031783 on Build GRCh38
rs755409657 Chr.7: 101031752-101031752 on Build GRCh38
rs537936932 Chr.7: 101031916-101031916 on Build GRCh38
rs748284769 Chr.7: 101031763-101031763 on Build GRCh38
rs370094020 Chr.7: 101031820-101031820 on Build GRCh38
rs542185793 Chr.7: 101031871-101031871 on Build GRCh38
rs753164036 Chr.7: 101031957-101031957 on Build GRCh38
rs745709640 Chr.7: 101031867-101031867 on Build GRCh38
rs200331855 Chr.7: 101031878-101031878 on Build GRCh38
rs866756152 Chr.7: 101031942-101031942 on Build GRCh38
rs140150533 Chr.7: 101031742-101031742 on Build GRCh38
rs767825541 Chr.7: 101031926-101031926 on Build GRCh38
rs866664184 Chr.7: 101031846-101031846 on Build GRCh38
rs755321879 Chr.7: 101031835-101031835 on Build GRCh38
rs371869918 Chr.7: 101031852-101031852 on Build GRCh38
rs199776258 Chr.7: 101031894-101031894 on Build GRCh38
rs867126570 Chr.7: 101031829-101031829 on Build GRCh38
rs866818830 Chr.7: 101031799-101031799 on Build GRCh38
rs745804040 Chr.7: 101031743-101031743 on Build GRCh38
rs369222127 Chr.7: 101031897-101031897 on Build GRCh38
rs1011437157 Chr.7: 101031791-101031791 on Build GRCh38
rs756193485 Chr.7: 101031766-101031766 on Build GRCh38
rs375199666 Chr.7: 101031771-101031771 on Build GRCh38
rs769261570 Chr.7: 101031854-101031855 on Build GRCh38
rs974330215 Chr.7: 101031918-101031918 on Build GRCh38
rs1023233712 Chr.7: 101031794-101031794 on Build GRCh38
rs749483762 Chr.7: 101031775-101031775 on Build GRCh38
rs775029580 Chr.7: 101031861-101031861 on Build GRCh38
rs779269229 Chr.7: 101031859-101031859 on Build GRCh38
rs751987235 Chr.7: 101031834-101031834 on Build GRCh38
rs567787391 Chr.7: 101031900-101031900 on Build GRCh38
rs111227419 Chr.7: 101031883-101031883 on Build GRCh38
rs777857466 Chr.7: 101031853-101031853 on Build GRCh38
rs1051384198 Chr.7: 101031739-101031739 on Build GRCh38
rs115500432 Chr.7: 101031851-101031851 on Build GRCh38
rs770648410 Chr.7: 101031778-101031778 on Build GRCh38
rs375249312 Chr.7: 101031854-101031854 on Build GRCh38
rs76440115 Chr.7: 101031847-101031847 on Build GRCh38
rs567129894 Chr.7: 101031855-101031855 on Build GRCh38
rs151280870 Chr.7: 101031801-101031801 on Build GRCh38
rs760144084 Chr.7: 101031812-101031812 on Build GRCh38
rs774224446 Chr.7: 101031793-101031793 on Build GRCh38
rs764926409 Chr.7: 101031913-101031913 on Build GRCh38
rs763319222 Chr.7: 101031869-101031870 on Build GRCh38

View Full Product Details