The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024226.1 | NP_001019397.1 | 5 - 5 | 1292 | 122 |
NM_001024227.1 | NP_001019398.1 | 5 - 5 | 1326 | 122 | |
NM_001024228.1 | NP_001019399.1 | 5 - 5 | 1304 | 122 | |
NM_001658.3 | NP_001649.1 | 5 - 5 | 1207 | 122 | |
XM_017001233.1 | XP_016856722.1 | 4 - 4 | 1295 | 122 | |
GenBank mRNA | AF052179.1 | - | 5 - 5 | 1139 | 122 |
AF055002.1 | - | 5 - 5 | 1135 | 122 | |
AK000620.1 | - | 5 - 5 | 1207 | 122 | |
AK057838.1 | - | 5 - 5 | 1135 | 122 | |
AK057875.1 | - | 5 - 5 | 1145 | 122 | |
AK130737.1 | - | 5 - 5 | 1136 | 122 | |
AK293553.1 | - | 7 - 7 | 1102 | 122 | |
AK296020.1 | - | 5 - 5 | 1055 | 122 | |
AK297792.1 | - | 4 - 4 | 1117 | 122 | |
BC009247.2 | - | 5 - 5 | 1119 | 122 | |
BC010429.1 | - | 5 - 5 | 1132 | 122 | |
BC011358.1 | - | 5 - 5 | 1158 | 122 | |
M36340.1 | - | 5 - 5 | 1139 | 122 | |
M84326.1 | - | 5 - 5 | 1136 | 122 |