Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 173917114-173917367 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SERPINC1
Entrez Gene ID
462
Gene Name
serpin family C member 1
Gene Aliases
AT3, AT3D, ATIII, THPH7
UniGene
Hs.75599
Gene Chromosome Location
Chr.1: 173903804-173917378 on Build GRCh38
Transcripts
NM_000488.3 XM_005245198.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7167010 c.35G>C p.G12A Chr.1: 173917225-173917225 on Build GRCh38
4025228 c.25G>T p.V9L Chr.1: 173917235-173917235 on Build GRCh38
4597996 c.13G>A p.V5M Chr.1: 173917247-173917247 on Build GRCh38
1689223 c.20G>A p.G7E Chr.1: 173917240-173917240 on Build GRCh38
4896028 c.8C>T p.S3F Chr.1: 173917252-173917252 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs926046378 Chr.1: 173917268-173917268 on Build GRCh38
rs764606323 Chr.1: 173917306-173917306 on Build GRCh38
rs929133799 Chr.1: 173917326-173917326 on Build GRCh38
rs760952234 Chr.1: 173917178-173917178 on Build GRCh38
rs199469508 Chr.1: 173917217-173917217 on Build GRCh38
rs753196429 Chr.1: 173917197-173917197 on Build GRCh38
rs774577561 Chr.1: 173917145-173917145 on Build GRCh38
rs770098298 Chr.1: 173917285-173917285 on Build GRCh38
rs753713846 Chr.1: 173917243-173917243 on Build GRCh38
rs967432692 Chr.1: 173917232-173917232 on Build GRCh38
rs370576783 Chr.1: 173917263-173917263 on Build GRCh38
rs775919922 Chr.1: 173917290-173917290 on Build GRCh38
rs776992580 Chr.1: 173917184-173917184 on Build GRCh38
rs373264710 Chr.1: 173917331-173917331 on Build GRCh38
rs765797202 Chr.1: 173917195-173917195 on Build GRCh38
rs959235353 Chr.1: 173917209-173917209 on Build GRCh38
rs1038216197 Chr.1: 173917139-173917139 on Build GRCh38
rs777418425 Chr.1: 173917269-173917271 on Build GRCh38
rs375773615 Chr.1: 173917181-173917181 on Build GRCh38
rs776785547 Chr.1: 173917267-173917267 on Build GRCh38
rs778745435 Chr.1: 173917245-173917245 on Build GRCh38
rs200460215 Chr.1: 173917294-173917294 on Build GRCh38
rs772040776 Chr.1: 173917248-173917248 on Build GRCh38
rs202085376 Chr.1: 173917211-173917211 on Build GRCh38
rs483352843 Chr.1: 173917270-173917270 on Build GRCh38
rs777886588 Chr.1: 173917257-173917257 on Build GRCh38
rs896860865 Chr.1: 173917164-173917164 on Build GRCh38
rs61736655 Chr.1: 173917231-173917231 on Build GRCh38
rs1001880914 Chr.1: 173917165-173917165 on Build GRCh38
rs887543419 Chr.1: 173917343-173917343 on Build GRCh38
rs1040206481 Chr.1: 173917307-173917307 on Build GRCh38
rs750709130 Chr.1: 173917247-173917247 on Build GRCh38
rs748072934 Chr.1: 173917246-173917246 on Build GRCh38
rs1020454535 Chr.1: 173917265-173917265 on Build GRCh38
rs372524963 Chr.1: 173917316-173917316 on Build GRCh38
rs759854656 Chr.1: 173917190-173917190 on Build GRCh38
rs771146323 Chr.1: 173917264-173917264 on Build GRCh38
rs767005471 Chr.1: 173917207-173917207 on Build GRCh38
rs1051194995 Chr.1: 173917170-173917170 on Build GRCh38
rs376206880 Chr.1: 173917347-173917347 on Build GRCh38
rs887409510 Chr.1: 173917146-173917146 on Build GRCh38
rs981581302 Chr.1: 173917293-173917293 on Build GRCh38
rs779790099 Chr.1: 173917235-173917235 on Build GRCh38
rs199469506 Chr.1: 173917254-173917254 on Build GRCh38
rs763486422 Chr.1: 173917198-173917198 on Build GRCh38

View Full Product Details