Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 69248141-69248402 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCA5
Entrez Gene ID
23461
Gene Name
ATP binding cassette subfamily A member 5
Gene Aliases
ABC13, EST90625
UniGene
Hs.731824
Gene Chromosome Location
Chr.17: 69244435-69327182 on Build GRCh38
Transcripts
NM_018672.4 NM_172232.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2975692 c.4766-1G>T p.? Chr.17: 69248318-69248318 on Build GRCh38
344588 c.4769A>T p.K1590I Chr.17: 69248314-69248314 on Build GRCh38
6365592 c.4808C>G p.A1603G Chr.17: 69248275-69248275 on Build GRCh38
6249648 c.4782C>A p.A1594A Chr.17: 69248301-69248301 on Build GRCh38
6569575 c.4821+3A>T p.? Chr.17: 69248259-69248259 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776415905 Chr.17: 69248193-69248193 on Build GRCh38
rs750280986 Chr.17: 69248270-69248270 on Build GRCh38
rs765222882 Chr.17: 69248269-69248269 on Build GRCh38
rs929088322 Chr.17: 69248207-69248207 on Build GRCh38
rs780693507 Chr.17: 69248292-69248292 on Build GRCh38
rs750100724 Chr.17: 69248359-69248359 on Build GRCh38
rs747608611 Chr.17: 69248296-69248296 on Build GRCh38
rs754519947 Chr.17: 69248287-69248287 on Build GRCh38
rs573795896 Chr.17: 69248267-69248267 on Build GRCh38
rs772368177 Chr.17: 69248345-69248345 on Build GRCh38
rs751218619 Chr.17: 69248276-69248276 on Build GRCh38
rs147523276 Chr.17: 69248300-69248300 on Build GRCh38
rs774507471 Chr.17: 69248335-69248335 on Build GRCh38
rs775739121 Chr.17: 69248346-69248346 on Build GRCh38
rs573479332 Chr.17: 69248174-69248174 on Build GRCh38
rs767634576 Chr.17: 69248256-69248256 on Build GRCh38
rs1046693327 Chr.17: 69248228-69248228 on Build GRCh38
rs369409961 Chr.17: 69248320-69248320 on Build GRCh38
rs910582292 Chr.17: 69248187-69248187 on Build GRCh38
rs1042088444 Chr.17: 69248374-69248374 on Build GRCh38
rs953408164 Chr.17: 69248226-69248226 on Build GRCh38
rs775639651 Chr.17: 69248309-69248309 on Build GRCh38
rs533978219 Chr.17: 69248182-69248182 on Build GRCh38
rs759594562 Chr.17: 69248250-69248250 on Build GRCh38
rs749666297 Chr.17: 69248312-69248312 on Build GRCh38
rs752761038 Chr.17: 69248261-69248261 on Build GRCh38
rs866503362 Chr.17: 69248302-69248302 on Build GRCh38
rs569280653 Chr.17: 69248179-69248179 on Build GRCh38
rs199719094 Chr.17: 69248349-69248349 on Build GRCh38
rs763338995 Chr.17: 69248227-69248227 on Build GRCh38
rs973460420 Chr.17: 69248200-69248200 on Build GRCh38
rs927900635 Chr.17: 69248229-69248229 on Build GRCh38
rs774613336 Chr.17: 69248240-69248240 on Build GRCh38
rs563156143 Chr.17: 69248351-69248351 on Build GRCh38
rs778223624 Chr.17: 69248306-69248306 on Build GRCh38
rs377045955 Chr.17: 69248206-69248206 on Build GRCh38
rs372859331 Chr.17: 69248333-69248333 on Build GRCh38
rs942117941 Chr.17: 69248315-69248318 on Build GRCh38
rs762536869 Chr.17: 69248358-69248359 on Build GRCh38
rs766199750 Chr.17: 69248368-69248368 on Build GRCh38
rs759412787 Chr.17: 69248275-69248275 on Build GRCh38
rs963383913 Chr.17: 69248196-69248196 on Build GRCh38
rs771100109 Chr.17: 69248238-69248238 on Build GRCh38
rs376105466 Chr.17: 69248352-69248352 on Build GRCh38
rs746111452 Chr.17: 69248343-69248343 on Build GRCh38

View Full Product Details