Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 75204090-75204363 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTRB2
Entrez Gene ID
440387
Gene Name
chymotrypsinogen B2
Gene Aliases
-
UniGene
Hs.610926
Gene Chromosome Location
Chr.16: 75204096-75207174 on Build GRCh38
Transcripts
NM_001025200.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4141903 c.654C>A p.V218V Chr.16: 75204299-75204299 on Build GRCh38
4558539 c.771G>A p.Q257Q Chr.16: 75204182-75204182 on Build GRCh38
4556180 c.681G>A p.L227L Chr.16: 75204272-75204272 on Build GRCh38
4825593 c.713G>A p.C238Y Chr.16: 75204240-75204240 on Build GRCh38
4579264 c.645C>A p.G215G Chr.16: 75204308-75204308 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755162448 Chr.16: 75204186-75204186 on Build GRCh38
rs202142318 Chr.16: 75204237-75204237 on Build GRCh38
rs748249913 Chr.16: 75204193-75204193 on Build GRCh38
rs964831894 Chr.16: 75204259-75204259 on Build GRCh38
rs976231541 Chr.16: 75204261-75204261 on Build GRCh38
rs978345244 Chr.16: 75204290-75204290 on Build GRCh38
rs774947076 Chr.16: 75204223-75204223 on Build GRCh38
rs772742144 Chr.16: 75204313-75204313 on Build GRCh38
rs373214805 Chr.16: 75204111-75204111 on Build GRCh38
rs896968146 Chr.16: 75204338-75204338 on Build GRCh38
rs767484270 Chr.16: 75204230-75204230 on Build GRCh38
rs200914367 Chr.16: 75204308-75204308 on Build GRCh38
rs368123078 Chr.16: 75204343-75204343 on Build GRCh38
rs369340318 Chr.16: 75204341-75204341 on Build GRCh38
rs372637324 Chr.16: 75204297-75204297 on Build GRCh38
rs376020915 Chr.16: 75204228-75204228 on Build GRCh38
rs769985585 Chr.16: 75204249-75204249 on Build GRCh38
rs760041473 Chr.16: 75204214-75204214 on Build GRCh38
rs771612252 Chr.16: 75204211-75204211 on Build GRCh38
rs1130511 Chr.16: 75204311-75204311 on Build GRCh38
rs199561534 Chr.16: 75204245-75204245 on Build GRCh38
rs867994359 Chr.16: 75204241-75204241 on Build GRCh38
rs750306978 Chr.16: 75204282-75204282 on Build GRCh38
rs570589360 Chr.16: 75204150-75204150 on Build GRCh38
rs766377326 Chr.16: 75204315-75204315 on Build GRCh38
rs200309186 Chr.16: 75204178-75204178 on Build GRCh38
rs777101874 Chr.16: 75204212-75204212 on Build GRCh38
rs1049298 Chr.16: 75204236-75204236 on Build GRCh38
rs758515615 Chr.16: 75204294-75204294 on Build GRCh38
rs897064492 Chr.16: 75204176-75204176 on Build GRCh38
rs762461730 Chr.16: 75204251-75204251 on Build GRCh38
rs771506528 Chr.16: 75204312-75204312 on Build GRCh38
rs369741106 Chr.16: 75204115-75204115 on Build GRCh38
rs150936688 Chr.16: 75204224-75204224 on Build GRCh38
rs779071927 Chr.16: 75204191-75204191 on Build GRCh38
rs150266828 Chr.16: 75204264-75204264 on Build GRCh38
rs746390962 Chr.16: 75204247-75204247 on Build GRCh38
rs201246675 Chr.16: 75204147-75204147 on Build GRCh38
rs768405626 Chr.16: 75204252-75204252 on Build GRCh38
rs375207313 Chr.16: 75204337-75204337 on Build GRCh38
rs201069805 Chr.16: 75204246-75204246 on Build GRCh38
rs780969316 Chr.16: 75204114-75204114 on Build GRCh38
rs752798185 Chr.16: 75204243-75204243 on Build GRCh38
rs748217903 Chr.16: 75204144-75204144 on Build GRCh38
rs1130512 Chr.16: 75204293-75204293 on Build GRCh38
rs751763983 Chr.16: 75204333-75204333 on Build GRCh38
rs763672343 Chr.16: 75204165-75204165 on Build GRCh38
rs1049303 Chr.16: 75204170-75204170 on Build GRCh38
rs759239567 Chr.16: 75204156-75204156 on Build GRCh38
rs899622435 Chr.16: 75204328-75204328 on Build GRCh38
rs376408035 Chr.16: 75204112-75204112 on Build GRCh38
rs747480822 Chr.16: 75204210-75204210 on Build GRCh38
rs1130509 Chr.16: 75204320-75204320 on Build GRCh38
rs4737 Chr.16: 75204205-75204205 on Build GRCh38
rs138352937 Chr.16: 75204304-75204304 on Build GRCh38
rs750268294 Chr.16: 75204180-75204180 on Build GRCh38
rs370422301 Chr.16: 75204142-75204142 on Build GRCh38
rs368315546 Chr.16: 75204286-75204286 on Build GRCh38
rs903490883 Chr.16: 75204131-75204131 on Build GRCh38
rs1049299 Chr.16: 75204232-75204232 on Build GRCh38
rs764474908 Chr.16: 75204295-75204295 on Build GRCh38
rs767429128 Chr.16: 75204278-75204278 on Build GRCh38
rs143963707 Chr.16: 75204215-75204215 on Build GRCh38
rs760483646 Chr.16: 75204314-75204314 on Build GRCh38
rs771991353 Chr.16: 75204197-75204197 on Build GRCh38
rs756664427 Chr.16: 75204301-75204301 on Build GRCh38
rs34824152 Chr.16: 75204164-75204164 on Build GRCh38
rs1042434789 Chr.16: 75204118-75204118 on Build GRCh38
rs1047024446 Chr.16: 75204305-75204305 on Build GRCh38
rs373513883 Chr.16: 75204117-75204117 on Build GRCh38
rs373944434 Chr.16: 75204154-75204154 on Build GRCh38
rs200581285 Chr.16: 75204157-75204157 on Build GRCh38
rs137960796 Chr.16: 75204250-75204251 on Build GRCh38
rs755921589 Chr.16: 75204234-75204234 on Build GRCh38
rs780257400 Chr.16: 75204302-75204302 on Build GRCh38
rs367685599 Chr.16: 75204326-75204326 on Build GRCh38
rs781425687 Chr.16: 75204299-75204299 on Build GRCh38
rs759430138 Chr.16: 75204322-75204322 on Build GRCh38
rs995167810 Chr.16: 75204145-75204145 on Build GRCh38
rs751430556 Chr.16: 75204169-75204169 on Build GRCh38
rs371932307 Chr.16: 75204335-75204335 on Build GRCh38
rs764712699 Chr.16: 75204119-75204119 on Build GRCh38
rs147088344 Chr.16: 75204296-75204296 on Build GRCh38
rs1049293 Chr.16: 75204287-75204287 on Build GRCh38
rs775952669 Chr.16: 75204250-75204250 on Build GRCh38

View Full Product Details