Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 2250902-2251127 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMH
Entrez Gene ID
268
Gene Name
anti-Mullerian hormone
Gene Aliases
MIF, MIS
UniGene
Hs.112432
Gene Chromosome Location
Chr.19: 2249114-2252073 on Build GRCh38
Transcripts
NM_000479.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5434681 c.841G>C p.E281Q Chr.19: 2251115-2251115 on Build GRCh38
4131577 c.791G>T p.G264V Chr.19: 2250975-2250975 on Build GRCh38
5474123 c.719G>A p.R240H Chr.19: 2250903-2250903 on Build GRCh38
6456096 c.758C>T p.P253L Chr.19: 2250942-2250942 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs749781953 Chr.19: 2251004-2251004 on Build GRCh38
rs779568794 Chr.19: 2251097-2251097 on Build GRCh38
rs754795092 Chr.19: 2251076-2251076 on Build GRCh38
rs376247795 Chr.19: 2250974-2250974 on Build GRCh38
rs762823966 Chr.19: 2251060-2251060 on Build GRCh38
rs774896174 Chr.19: 2250936-2250936 on Build GRCh38
rs757506343 Chr.19: 2250992-2250992 on Build GRCh38
rs766314318 Chr.19: 2251069-2251069 on Build GRCh38
rs753231264 Chr.19: 2250945-2250945 on Build GRCh38
rs763830481 Chr.19: 2250943-2250943 on Build GRCh38
rs756330898 Chr.19: 2251080-2251080 on Build GRCh38
rs776344517 Chr.19: 2251031-2251031 on Build GRCh38
rs749401185 Chr.19: 2251087-2251087 on Build GRCh38
rs746250086 Chr.19: 2251020-2251020 on Build GRCh38
rs752939885 Chr.19: 2251077-2251077 on Build GRCh38
rs565488272 Chr.19: 2251106-2251106 on Build GRCh38
rs1019871219 Chr.19: 2250949-2250949 on Build GRCh38
rs546849156 Chr.19: 2250998-2250998 on Build GRCh38
rs764785266 Chr.19: 2251045-2251045 on Build GRCh38
rs973412835 Chr.19: 2251044-2251044 on Build GRCh38
rs1038800744 Chr.19: 2250968-2250968 on Build GRCh38
rs750351723 Chr.19: 2250950-2250950 on Build GRCh38
rs201779218 Chr.19: 2251108-2251108 on Build GRCh38
rs778047263 Chr.19: 2251001-2251001 on Build GRCh38
rs991688114 Chr.19: 2251099-2251099 on Build GRCh38
rs981817930 Chr.19: 2251024-2251024 on Build GRCh38
rs774808404 Chr.19: 2251018-2251018 on Build GRCh38
rs772753956 Chr.19: 2251052-2251052 on Build GRCh38
rs1008450940 Chr.19: 2251028-2251028 on Build GRCh38
rs758383760 Chr.19: 2250973-2250973 on Build GRCh38
rs761433484 Chr.19: 2251042-2251042 on Build GRCh38
rs760364562 Chr.19: 2250942-2250942 on Build GRCh38
rs771597672 Chr.19: 2251005-2251005 on Build GRCh38
rs768128327 Chr.19: 2251027-2251027 on Build GRCh38
rs781479377 Chr.19: 2250991-2250991 on Build GRCh38
rs371054070 Chr.19: 2251089-2251089 on Build GRCh38
rs751285539 Chr.19: 2251074-2251074 on Build GRCh38
rs764724666 Chr.19: 2250946-2250946 on Build GRCh38
rs746431360 Chr.19: 2251102-2251102 on Build GRCh38
rs201324658 Chr.19: 2250986-2250986 on Build GRCh38
rs949969959 Chr.19: 2250926-2250926 on Build GRCh38
rs940175165 Chr.19: 2251032-2251032 on Build GRCh38

View Full Product Details