Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 167821949-167822214 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADCY10
Entrez Gene ID
55811
Gene Name
adenylate cyclase 10, soluble
Gene Aliases
HCA2, HEL-S-7a, SAC, SACI, Sacy, hsAC
UniGene
Hs.320892
Gene Chromosome Location
Chr.1: 167809080-167915556 on Build GRCh38
Transcripts
NM_001167749.2 NM_001297772.1 NM_018417.5 XM_006711449.3 XM_011509760.2 XM_011509762.2 XM_011509763.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6594856 c.4240T>G p.F1414V Chr.1: 167822070-167822070 on Build GRCh38
458423 c.4189G>C p.E1397Q Chr.1: 167822121-167822121 on Build GRCh38
1244449 c.4218C>T p.Y1406Y Chr.1: 167822092-167822092 on Build GRCh38
5694522 c.4223A>G p.N1408S Chr.1: 167822087-167822087 on Build GRCh38
5983651 c.4181G>C p.R1394T Chr.1: 167822129-167822129 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781277868 Chr.1: 167822094-167822094 on Build GRCh38
rs61737676 Chr.1: 167822090-167822090 on Build GRCh38
rs776903771 Chr.1: 167822017-167822017 on Build GRCh38
rs774417777 Chr.1: 167822135-167822135 on Build GRCh38
rs768796489 Chr.1: 167822016-167822016 on Build GRCh38
rs150684208 Chr.1: 167822131-167822131 on Build GRCh38
rs1035911834 Chr.1: 167822054-167822054 on Build GRCh38
rs752389676 Chr.1: 167822060-167822060 on Build GRCh38
rs765104099 Chr.1: 167822109-167822109 on Build GRCh38
rs201132987 Chr.1: 167822058-167822058 on Build GRCh38
rs775016525 Chr.1: 167822155-167822155 on Build GRCh38
rs748246675 Chr.1: 167822020-167822020 on Build GRCh38
rs945499325 Chr.1: 167822024-167822024 on Build GRCh38
rs755314315 Chr.1: 167822183-167822183 on Build GRCh38
rs1015945868 Chr.1: 167822126-167822126 on Build GRCh38
rs555126677 Chr.1: 167822182-167822182 on Build GRCh38
rs200525715 Chr.1: 167822091-167822091 on Build GRCh38
rs200037968 Chr.1: 167822034-167822034 on Build GRCh38
rs763534282 Chr.1: 167822077-167822077 on Build GRCh38
rs901346962 Chr.1: 167822063-167822063 on Build GRCh38
rs774328227 Chr.1: 167822045-167822045 on Build GRCh38
rs760323146 Chr.1: 167822158-167822158 on Build GRCh38
rs773297917 Chr.1: 167822023-167822023 on Build GRCh38
rs781573590 Chr.1: 167822187-167822187 on Build GRCh38
rs763621713 Chr.1: 167822163-167822163 on Build GRCh38
rs748350987 Chr.1: 167822188-167822188 on Build GRCh38
rs148081594 Chr.1: 167822098-167822098 on Build GRCh38
rs201700580 Chr.1: 167822036-167822036 on Build GRCh38
rs371891859 Chr.1: 167821992-167821992 on Build GRCh38
rs755582452 Chr.1: 167822066-167822066 on Build GRCh38
rs780344205 Chr.1: 167822008-167822008 on Build GRCh38
rs184493679 Chr.1: 167822136-167822136 on Build GRCh38
rs748797897 Chr.1: 167821984-167821984 on Build GRCh38
rs769788816 Chr.1: 167822105-167822105 on Build GRCh38
rs779354653 Chr.1: 167821982-167821982 on Build GRCh38
rs756733779 Chr.1: 167822170-167822170 on Build GRCh38
rs993264564 Chr.1: 167822184-167822184 on Build GRCh38
rs753382284 Chr.1: 167822167-167822167 on Build GRCh38
rs761396509 Chr.1: 167822144-167822145 on Build GRCh38
rs755370222 Chr.1: 167822092-167822092 on Build GRCh38
rs550447018 Chr.1: 167822012-167822012 on Build GRCh38
rs758859163 Chr.1: 167822083-167822083 on Build GRCh38
rs375622466 Chr.1: 167822078-167822078 on Build GRCh38
rs766942582 Chr.1: 167822171-167822171 on Build GRCh38
rs769847813 Chr.1: 167822021-167822021 on Build GRCh38
rs759352864 Chr.1: 167822057-167822057 on Build GRCh38
rs755636548 Chr.1: 167821980-167821980 on Build GRCh38
rs749349689 Chr.1: 167822124-167822124 on Build GRCh38
rs767274758 Chr.1: 167822148-167822148 on Build GRCh38

View Full Product Details