Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 43889636-43889909 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALKBH3
Entrez Gene ID
221120
Gene Name
alkB homolog 3, alpha-ketoglutaratedependent dioxygenase
Gene Aliases
ABH3, DEPC-1, DEPC1, PCA1, hABH3
UniGene
Hs.720708
Gene Chromosome Location
Chr.11: 43880807-43920275 on Build GRCh38
Transcripts
NM_139178.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4661145 c.284G>T p.G95V Chr.11: 43889742-43889742 on Build GRCh38
4032783 c.293T>C p.V98A Chr.11: 43889754-43889754 on Build GRCh38
4191618 c.273G>T p.L91F Chr.11: 43889734-43889734 on Build GRCh38
6503051 c.328C>G p.Q110E Chr.11: 43889789-43889789 on Build GRCh38
6503050 c.331C>G p.Q111E Chr.11: 43889789-43889789 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774357027 Chr.11: 43889880-43889880 on Build GRCh38
rs767771210 Chr.11: 43889788-43889788 on Build GRCh38
rs778328022 Chr.11: 43889816-43889816 on Build GRCh38
rs367974114 Chr.11: 43889689-43889689 on Build GRCh38
rs773338017 Chr.11: 43889681-43889681 on Build GRCh38
rs938332661 Chr.11: 43889808-43889808 on Build GRCh38
rs779698624 Chr.11: 43889849-43889849 on Build GRCh38
rs759963526 Chr.11: 43889691-43889691 on Build GRCh38
rs770864815 Chr.11: 43889756-43889756 on Build GRCh38
rs778370105 Chr.11: 43889704-43889704 on Build GRCh38
rs762961303 Chr.11: 43889687-43889687 on Build GRCh38
rs2434478 Chr.11: 43889815-43889815 on Build GRCh38
rs751689677 Chr.11: 43889690-43889690 on Build GRCh38
rs35076527 Chr.11: 43889661-43889661 on Build GRCh38
rs756747038 Chr.11: 43889703-43889703 on Build GRCh38
rs752895583 Chr.11: 43889808-43889809 on Build GRCh38
rs751630340 Chr.11: 43889793-43889793 on Build GRCh38
rs756790030 Chr.11: 43889708-43889708 on Build GRCh38
rs774464321 Chr.11: 43889765-43889765 on Build GRCh38
rs375599351 Chr.11: 43889717-43889717 on Build GRCh38
rs368878641 Chr.11: 43889821-43889821 on Build GRCh38
rs889817964 Chr.11: 43889882-43889882 on Build GRCh38
rs759713035 Chr.11: 43889766-43889766 on Build GRCh38
rs542847003 Chr.11: 43889748-43889748 on Build GRCh38
rs141900065 Chr.11: 43889671-43889671 on Build GRCh38
rs767816956 Chr.11: 43889698-43889698 on Build GRCh38
rs753112183 Chr.11: 43889700-43889700 on Build GRCh38
rs771725822 Chr.11: 43889728-43889728 on Build GRCh38
rs764732289 Chr.11: 43889799-43889799 on Build GRCh38
rs758043229 Chr.11: 43889835-43889835 on Build GRCh38
rs776482589 Chr.11: 43889752-43889752 on Build GRCh38
rs200278238 Chr.11: 43889753-43889753 on Build GRCh38
rs528763655 Chr.11: 43889862-43889862 on Build GRCh38
rs753128786 Chr.11: 43889791-43889791 on Build GRCh38
rs765398799 Chr.11: 43889693-43889699 on Build GRCh38
rs186623067 Chr.11: 43889870-43889870 on Build GRCh38
rs748129614 Chr.11: 43889871-43889871 on Build GRCh38
rs972939647 Chr.11: 43889789-43889789 on Build GRCh38
rs779780686 Chr.11: 43889741-43889741 on Build GRCh38
rs759498852 Chr.11: 43889822-43889823 on Build GRCh38
rs200004652 Chr.11: 43889863-43889863 on Build GRCh38
rs745557232 Chr.11: 43889725-43889725 on Build GRCh38
rs143155140 Chr.11: 43889800-43889800 on Build GRCh38
rs150554870 Chr.11: 43889706-43889706 on Build GRCh38
rs761057713 Chr.11: 43889797-43889797 on Build GRCh38
rs532665667 Chr.11: 43889876-43889876 on Build GRCh38

View Full Product Details