Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 418713-418942 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANO9
Entrez Gene ID
338440
Gene Name
anoctamin 9
Gene Aliases
PIG5, TMEM16J, TP53I5
UniGene
Hs.501622
Gene Chromosome Location
Chr.11: 417930-442011 on Build GRCh38
Transcripts
NM_001012302.2 XM_011520053.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5779334 c.2119A>G p.I707V Chr.11: 418731-418731 on Build GRCh38
7163485 c.2008G>C p.E670Q Chr.11: 418916-418916 on Build GRCh38
926865 c.2062G>A p.D688N Chr.11: 418788-418788 on Build GRCh38
6601208 c.2051G>A p.R684H Chr.11: 418799-418799 on Build GRCh38
4662088 c.2029_2030delCT p.L677fs*33 Chr.11: 418894-418895 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs576751915 Chr.11: 418843-418843 on Build GRCh38
rs753013286 Chr.11: 418800-418800 on Build GRCh38
rs757856230 Chr.11: 418832-418832 on Build GRCh38
rs765892541 Chr.11: 418894-418895 on Build GRCh38
rs563085071 Chr.11: 418798-418798 on Build GRCh38
rs771565337 Chr.11: 418878-418878 on Build GRCh38
rs762839755 Chr.11: 418774-418774 on Build GRCh38
rs367921445 Chr.11: 418901-418901 on Build GRCh38
rs777693518 Chr.11: 418750-418750 on Build GRCh38
rs201214547 Chr.11: 418792-418792 on Build GRCh38
rs769338224 Chr.11: 418773-418773 on Build GRCh38
rs773861579 Chr.11: 418865-418865 on Build GRCh38
rs371131273 Chr.11: 418856-418856 on Build GRCh38
rs777159075 Chr.11: 418770-418770 on Build GRCh38
rs116108490 Chr.11: 418745-418745 on Build GRCh38
rs758562981 Chr.11: 418746-418746 on Build GRCh38
rs746360214 Chr.11: 418838-418838 on Build GRCh38
rs373327405 Chr.11: 418799-418799 on Build GRCh38
rs764356229 Chr.11: 418923-418923 on Build GRCh38
rs371132860 Chr.11: 418789-418789 on Build GRCh38
rs757150834 Chr.11: 418734-418734 on Build GRCh38
rs759945469 Chr.11: 418830-418830 on Build GRCh38
rs778032728 Chr.11: 418817-418817 on Build GRCh38
rs747558548 Chr.11: 418758-418758 on Build GRCh38
rs758517780 Chr.11: 418810-418810 on Build GRCh38
rs781738375 Chr.11: 418837-418837 on Build GRCh38
rs374074944 Chr.11: 418869-418869 on Build GRCh38
rs371374296 Chr.11: 418902-418902 on Build GRCh38
rs760506226 Chr.11: 418897-418897 on Build GRCh38
rs755167156 Chr.11: 418740-418740 on Build GRCh38
rs763260469 Chr.11: 418908-418908 on Build GRCh38
rs771526322 Chr.11: 418763-418763 on Build GRCh38
rs750166102 Chr.11: 418739-418739 on Build GRCh38
rs368497607 Chr.11: 418848-418848 on Build GRCh38
rs759337830 Chr.11: 418828-418828 on Build GRCh38
rs773192364 Chr.11: 418890-418890 on Build GRCh38
rs377737999 Chr.11: 418788-418788 on Build GRCh38
rs12792208 Chr.11: 418735-418735 on Build GRCh38
rs193920739 Chr.11: 418804-418804 on Build GRCh38
rs925026417 Chr.11: 418791-418791 on Build GRCh38
rs746180111 Chr.11: 418771-418771 on Build GRCh38
rs762502699 Chr.11: 418786-418786 on Build GRCh38
rs767383937 Chr.11: 418793-418793 on Build GRCh38
rs777165017 Chr.11: 418806-418806 on Build GRCh38

View Full Product Details