Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 79834003-79834258 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CBX4
Entrez Gene ID
8535
Gene Name
chromobox 4
Gene Aliases
NBP16, PC2
UniGene
Hs.405046
Gene Chromosome Location
Chr.17: 79833156-79839414 on Build GRCh38
Transcripts
NM_003655.2 XM_011525399.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3388267 c.1439C>T p.T480M Chr.17: 79834203-79834203 on Build GRCh38
4400793 c.1421A>G p.D474G Chr.17: 79834221-79834221 on Build GRCh38
5387552 c.1638C>T p.T546T Chr.17: 79834004-79834004 on Build GRCh38
6854883 c.1539G>A p.T513T Chr.17: 79834103-79834103 on Build GRCh38
4467114 c.1470C>T p.S490S Chr.17: 79834172-79834172 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771101934 Chr.17: 79834091-79834091 on Build GRCh38
rs924884978 Chr.17: 79834139-79834144 on Build GRCh38
rs764827212 Chr.17: 79834125-79834125 on Build GRCh38
rs762730314 Chr.17: 79834178-79834178 on Build GRCh38
rs749940879 Chr.17: 79834049-79834049 on Build GRCh38
rs772135793 Chr.17: 79834152-79834152 on Build GRCh38
rs756548734 Chr.17: 79834075-79834075 on Build GRCh38
rs757568848 Chr.17: 79834200-79834200 on Build GRCh38
rs992599983 Chr.17: 79834037-79834037 on Build GRCh38
rs376832453 Chr.17: 79834131-79834131 on Build GRCh38
rs764759904 Chr.17: 79834122-79834123 on Build GRCh38
rs757906326 Chr.17: 79834134-79834135 on Build GRCh38
rs749975542 Chr.17: 79834127-79834128 on Build GRCh38
rs564210946 Chr.17: 79834072-79834072 on Build GRCh38
rs774452384 Chr.17: 79834098-79834098 on Build GRCh38
rs749648445 Chr.17: 79834142-79834142 on Build GRCh38
rs200965379 Chr.17: 79834128-79834128 on Build GRCh38
rs201414154 Chr.17: 79834157-79834157 on Build GRCh38
rs761409501 Chr.17: 79834123-79834128 on Build GRCh38
rs549712900 Chr.17: 79834100-79834100 on Build GRCh38
rs775410592 Chr.17: 79834104-79834104 on Build GRCh38
rs751160083 Chr.17: 79834189-79834189 on Build GRCh38
rs779018829 Chr.17: 79834146-79834146 on Build GRCh38
rs749503302 Chr.17: 79834090-79834090 on Build GRCh38
rs201780947 Chr.17: 79834140-79834140 on Build GRCh38
rs199722528 Chr.17: 79834169-79834169 on Build GRCh38
rs1040492308 Chr.17: 79834171-79834171 on Build GRCh38
rs761494208 Chr.17: 79834124-79834124 on Build GRCh38
rs760673997 Chr.17: 79834110-79834110 on Build GRCh38
rs761227308 Chr.17: 79834193-79834193 on Build GRCh38
rs765968311 Chr.17: 79834187-79834187 on Build GRCh38
rs745827861 Chr.17: 79834111-79834112 on Build GRCh38
rs763862805 Chr.17: 79834115-79834115 on Build GRCh38
rs146523753 Chr.17: 79834058-79834058 on Build GRCh38
rs780165123 Chr.17: 79834156-79834156 on Build GRCh38
rs754126884 Chr.17: 79834118-79834119 on Build GRCh38
rs779400926 Chr.17: 79834205-79834205 on Build GRCh38
rs900577226 Chr.17: 79834191-79834191 on Build GRCh38
rs781769596 Chr.17: 79834070-79834070 on Build GRCh38
rs764722519 Chr.17: 79834195-79834195 on Build GRCh38
rs1007896340 Chr.17: 79834077-79834077 on Build GRCh38
rs568219945 Chr.17: 79834127-79834127 on Build GRCh38
rs776459002 Chr.17: 79834118-79834118 on Build GRCh38
rs757454510 Chr.17: 79834145-79834145 on Build GRCh38
rs867588853 Chr.17: 79834081-79834081 on Build GRCh38
rs769427551 Chr.17: 79834165-79834165 on Build GRCh38
rs1021803587 Chr.17: 79834123-79834134 on Build GRCh38
rs565830961 Chr.17: 79834161-79834161 on Build GRCh38
rs12949594 Chr.17: 79834180-79834180 on Build GRCh38
rs776282365 Chr.17: 79834120-79834134 on Build GRCh38
rs753247490 Chr.17: 79834136-79834136 on Build GRCh38
rs908027065 Chr.17: 79834139-79834139 on Build GRCh38
rs758667165 Chr.17: 79834216-79834216 on Build GRCh38
rs916970832 Chr.17: 79834048-79834048 on Build GRCh38
rs372077305 Chr.17: 79834196-79834196 on Build GRCh38
rs771989725 Chr.17: 79834103-79834103 on Build GRCh38
rs529192897 Chr.17: 79834137-79834137 on Build GRCh38
rs531394505 Chr.17: 79834095-79834095 on Build GRCh38
rs778343571 Chr.17: 79834076-79834076 on Build GRCh38
rs750598235 Chr.17: 79834211-79834211 on Build GRCh38
rs915873206 Chr.17: 79834159-79834159 on Build GRCh38
rs547706029 Chr.17: 79834148-79834148 on Build GRCh38

View Full Product Details