Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 104724198-104724447 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADSSL1
Entrez Gene ID
122622
Gene Name
adenylosuccinate synthase like 1
Gene Aliases
-
UniGene
Hs.592327
Gene Chromosome Location
Chr.14: 104724197-104747325 on Build GRCh38
Transcripts
NM_152328.4 XM_006720026.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6239957 c.108G>T p.V36V Chr.14: 104724378-104724378 on Build GRCh38
7332280 c.5C>T p.S2L Chr.14: 104724275-104724275 on Build GRCh38
6030680 c.81G>C p.A27A Chr.14: 104724351-104724351 on Build GRCh38
6431791 c.160G>A p.A54T Chr.14: 104724430-104724430 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs763589704 Chr.14: 104724397-104724397 on Build GRCh38
rs369883033 Chr.14: 104724389-104724389 on Build GRCh38
rs771061801 Chr.14: 104724385-104724385 on Build GRCh38
rs988752817 Chr.14: 104724218-104724219 on Build GRCh38
rs962261994 Chr.14: 104724222-104724222 on Build GRCh38
rs1007996082 Chr.14: 104724387-104724387 on Build GRCh38
rs920736921 Chr.14: 104724229-104724229 on Build GRCh38
rs930838323 Chr.14: 104724234-104724234 on Build GRCh38
rs914227583 Chr.14: 104724235-104724235 on Build GRCh38
rs374922910 Chr.14: 104724357-104724357 on Build GRCh38
rs1054789257 Chr.14: 104724343-104724343 on Build GRCh38
rs755867684 Chr.14: 104724330-104724330 on Build GRCh38
rs892963326 Chr.14: 104724345-104724345 on Build GRCh38
rs950040469 Chr.14: 104724243-104724243 on Build GRCh38
rs555319574 Chr.14: 104724279-104724279 on Build GRCh38
rs1010149993 Chr.14: 104724430-104724430 on Build GRCh38
rs202092105 Chr.14: 104724392-104724392 on Build GRCh38
rs780933927 Chr.14: 104724248-104724248 on Build GRCh38
rs938452100 Chr.14: 104724262-104724262 on Build GRCh38
rs746999644 Chr.14: 104724378-104724378 on Build GRCh38
rs996618149 Chr.14: 104724347-104724347 on Build GRCh38
rs749979584 Chr.14: 104724421-104724421 on Build GRCh38
rs972330135 Chr.14: 104724224-104724224 on Build GRCh38
rs558001982 Chr.14: 104724367-104724367 on Build GRCh38
rs972331395 Chr.14: 104724417-104724417 on Build GRCh38
rs570126709 Chr.14: 104724240-104724240 on Build GRCh38
rs756674137 Chr.14: 104724404-104724404 on Build GRCh38
rs763479592 Chr.14: 104724393-104724393 on Build GRCh38
rs781559734 Chr.14: 104724301-104724301 on Build GRCh38
rs779795870 Chr.14: 104724354-104724354 on Build GRCh38
rs765892435 Chr.14: 104724391-104724391 on Build GRCh38
rs751239230 Chr.14: 104724398-104724398 on Build GRCh38
rs535599676 Chr.14: 104724242-104724242 on Build GRCh38
rs543561314 Chr.14: 104724420-104724420 on Build GRCh38
rs780029026 Chr.14: 104724426-104724426 on Build GRCh38
rs947070620 Chr.14: 104724239-104724239 on Build GRCh38
rs903210548 Chr.14: 104724251-104724251 on Build GRCh38
rs572140747 Chr.14: 104724283-104724283 on Build GRCh38
rs745830815 Chr.14: 104724282-104724282 on Build GRCh38
rs997124765 Chr.14: 104724351-104724351 on Build GRCh38
rs756027182 Chr.14: 104724422-104724422 on Build GRCh38
rs1057476383 Chr.14: 104724263-104724263 on Build GRCh38
rs563375481 Chr.14: 104724427-104724427 on Build GRCh38

View Full Product Details