Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 30069170-30069411 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALDOA
Entrez Gene ID
226
Gene Name
aldolase, fructose-bisphosphate A
Gene Aliases
ALDA, GSD12, HEL-S-87p
UniGene
Hs.513490
Gene Chromosome Location
Chr.16: 30053090-30070420 on Build GRCh38
Transcripts
NM_000034.3 NM_001127617.2 NM_001243177.1 NM_184041.2 NM_184043.2 XM_011545768.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5507326 c.541-6C>T p.? Chr.16: 30069300-30069300 on Build GRCh38
1749550 c.568G>A p.E190K Chr.16: 30069333-30069333 on Build GRCh38
341066 c.577C>T p.P193S Chr.16: 30069342-30069342 on Build GRCh38
3111672 c.598A>T p.K200* Chr.16: 30069363-30069363 on Build GRCh38
1301851 c.580G>A p.D194N Chr.16: 30069345-30069345 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752478544 Chr.16: 30069272-30069272 on Build GRCh38
rs759847191 Chr.16: 30069365-30069365 on Build GRCh38
rs758724483 Chr.16: 30069289-30069290 on Build GRCh38
rs749215165 Chr.16: 30069278-30069278 on Build GRCh38
rs920058176 Chr.16: 30069229-30069229 on Build GRCh38
rs745528292 Chr.16: 30069294-30069294 on Build GRCh38
rs372394541 Chr.16: 30069366-30069366 on Build GRCh38
rs201141448 Chr.16: 30069270-30069270 on Build GRCh38
rs930382668 Chr.16: 30069254-30069254 on Build GRCh38
rs775117761 Chr.16: 30069299-30069299 on Build GRCh38
rs186785222 Chr.16: 30069297-30069297 on Build GRCh38
rs755937471 Chr.16: 30069274-30069274 on Build GRCh38
rs550680791 Chr.16: 30069196-30069196 on Build GRCh38
rs1048332210 Chr.16: 30069231-30069231 on Build GRCh38
rs777581671 Chr.16: 30069382-30069382 on Build GRCh38
rs766537451 Chr.16: 30069256-30069256 on Build GRCh38
rs3112557 Chr.16: 30069352-30069352 on Build GRCh38
rs751632844 Chr.16: 30069362-30069362 on Build GRCh38
rs375097754 Chr.16: 30069302-30069302 on Build GRCh38
rs754308973 Chr.16: 30069375-30069375 on Build GRCh38
rs3751869 Chr.16: 30069199-30069199 on Build GRCh38
rs536229013 Chr.16: 30069280-30069280 on Build GRCh38
rs763478106 Chr.16: 30069341-30069341 on Build GRCh38
rs121909534 Chr.16: 30069384-30069384 on Build GRCh38
rs777632110 Chr.16: 30069275-30069275 on Build GRCh38
rs534264493 Chr.16: 30069367-30069367 on Build GRCh38
rs750622363 Chr.16: 30069286-30069286 on Build GRCh38
rs369668963 Chr.16: 30069377-30069377 on Build GRCh38
rs201519764 Chr.16: 30069317-30069317 on Build GRCh38
rs892947296 Chr.16: 30069391-30069391 on Build GRCh38
rs1012775602 Chr.16: 30069222-30069222 on Build GRCh38
rs781147998 Chr.16: 30069271-30069271 on Build GRCh38
rs766590264 Chr.16: 30069354-30069354 on Build GRCh38
rs894348239 Chr.16: 30069207-30069207 on Build GRCh38
rs755913779 Chr.16: 30069379-30069379 on Build GRCh38
rs573162475 Chr.16: 30069240-30069240 on Build GRCh38
rs1038936301 Chr.16: 30069224-30069224 on Build GRCh38
rs573479181 Chr.16: 30069323-30069323 on Build GRCh38

View Full Product Details