Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 26525522-26525781 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANO3
Entrez Gene ID
63982
Gene Name
anoctamin 3
Gene Aliases
C11orf25, DYT23, DYT24, GENX-3947, TMEM16C
UniGene
Hs.577269
Gene Chromosome Location
Chr.11: 26189123-26663289 on Build GRCh38
Transcripts
NM_001313726.1 NM_001313727.1 NM_031418.3 XM_017018118.1 XM_017018119.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2113046 c.712G>T p.D238Y Chr.11: 26525654-26525654 on Build GRCh38
926067 c.710C>T p.T237I Chr.11: 26525652-26525652 on Build GRCh38
4450578 c.693-1G>A p.? Chr.11: 26525634-26525634 on Build GRCh38
6429215 c.737G>A p.R246K Chr.11: 26525679-26525679 on Build GRCh38
3943607 c.712G>C p.D238H Chr.11: 26525654-26525654 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs952811795 Chr.11: 26525749-26525749 on Build GRCh38
rs374952377 Chr.11: 26525580-26525580 on Build GRCh38
rs571009442 Chr.11: 26525572-26525572 on Build GRCh38
rs1007188185 Chr.11: 26525680-26525680 on Build GRCh38
rs148719252 Chr.11: 26525657-26525657 on Build GRCh38
rs753255476 Chr.11: 26525625-26525626 on Build GRCh38
rs745381740 Chr.11: 26525676-26525676 on Build GRCh38
rs1024348703 Chr.11: 26525635-26525635 on Build GRCh38
rs1005579484 Chr.11: 26525752-26525752 on Build GRCh38
rs750791750 Chr.11: 26525716-26525716 on Build GRCh38
rs765750103 Chr.11: 26525715-26525715 on Build GRCh38
rs994578579 Chr.11: 26525719-26525719 on Build GRCh38
rs751681001 Chr.11: 26525722-26525722 on Build GRCh38
rs948003986 Chr.11: 26525577-26525577 on Build GRCh38
rs374948346 Chr.11: 26525621-26525621 on Build GRCh38
rs146532706 Chr.11: 26525656-26525656 on Build GRCh38
rs758677432 Chr.11: 26525605-26525605 on Build GRCh38
rs369401544 Chr.11: 26525581-26525581 on Build GRCh38
rs771596676 Chr.11: 26525675-26525675 on Build GRCh38
rs927692869 Chr.11: 26525598-26525598 on Build GRCh38
rs865944430 Chr.11: 26525671-26525671 on Build GRCh38
rs760058159 Chr.11: 26525686-26525686 on Build GRCh38
rs886037882 Chr.11: 26525644-26525644 on Build GRCh38
rs754954444 Chr.11: 26525727-26525727 on Build GRCh38
rs916386992 Chr.11: 26525557-26525557 on Build GRCh38
rs988902329 Chr.11: 26525706-26525706 on Build GRCh38
rs957553085 Chr.11: 26525700-26525700 on Build GRCh38
rs940481089 Chr.11: 26525564-26525564 on Build GRCh38
rs764678739 Chr.11: 26525694-26525694 on Build GRCh38
rs759237031 Chr.11: 26525674-26525674 on Build GRCh38
rs750736746 Chr.11: 26525586-26525586 on Build GRCh38
rs775101623 Chr.11: 26525681-26525681 on Build GRCh38
rs779279662 Chr.11: 26525585-26525585 on Build GRCh38
rs373560761 Chr.11: 26525720-26525720 on Build GRCh38
rs758883241 Chr.11: 26525635-26525636 on Build GRCh38
rs754228082 Chr.11: 26525712-26525712 on Build GRCh38
rs770780762 Chr.11: 26525661-26525661 on Build GRCh38
rs762402896 Chr.11: 26525714-26525714 on Build GRCh38
rs747028399 Chr.11: 26525632-26525632 on Build GRCh38
rs1020277417 Chr.11: 26525693-26525693 on Build GRCh38
rs976236127 Chr.11: 26525751-26525751 on Build GRCh38
rs774120335 Chr.11: 26525672-26525672 on Build GRCh38
rs144792604 Chr.11: 26525646-26525646 on Build GRCh38

View Full Product Details