Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 99284312-99284545 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADH1A
Entrez Gene ID
124
Gene Name
alcohol dehydrogenase 1A (class I), alpha polypeptide
Gene Aliases
ADH1
UniGene
Hs.654433
Gene Chromosome Location
Chr.4: 99276366-99291028 on Build GRCh38
Transcripts
NM_000667.3 NR_037884.1

Gene Symbol
LOC100507053
Entrez Gene ID
100507053
Gene Name
uncharacterized LOC100507053
Gene Aliases
-
UniGene
Hs.635084
Gene Chromosome Location
Chr.4: 99088857-99301356 on Build GRCh38
Transcripts
NM_000667.3 NR_037884.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1633379 c.501A>G p.L167L Chr.4: 99284465-99284465 on Build GRCh38
3598545 c.456G>A p.V152V Chr.4: 99284510-99284510 on Build GRCh38
732153 c.496C>A p.P166T Chr.4: 99284470-99284470 on Build GRCh38
1049402 c.537T>A p.T179T Chr.4: 99284429-99284429 on Build GRCh38
3365429 c.461A>T p.D154V Chr.4: 99284505-99284505 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745449007 Chr.4: 99284492-99284492 on Build GRCh38
rs267599959 Chr.4: 99284440-99284440 on Build GRCh38
rs766574286 Chr.4: 99284455-99284455 on Build GRCh38
rs772361611 Chr.4: 99284414-99284414 on Build GRCh38
rs143808002 Chr.4: 99284516-99284516 on Build GRCh38
rs756307800 Chr.4: 99284351-99284351 on Build GRCh38
rs746667944 Chr.4: 99284482-99284482 on Build GRCh38
rs141160298 Chr.4: 99284368-99284368 on Build GRCh38
rs758178003 Chr.4: 99284481-99284481 on Build GRCh38
rs762122061 Chr.4: 99284427-99284427 on Build GRCh38
rs893697772 Chr.4: 99284346-99284346 on Build GRCh38
rs771897654 Chr.4: 99284383-99284383 on Build GRCh38
rs915864673 Chr.4: 99284340-99284340 on Build GRCh38
rs377688831 Chr.4: 99284391-99284391 on Build GRCh38
rs116276567 Chr.4: 99284484-99284484 on Build GRCh38
rs746653344 Chr.4: 99284390-99284390 on Build GRCh38
rs755100175 Chr.4: 99284471-99284471 on Build GRCh38
rs774913626 Chr.4: 99284409-99284409 on Build GRCh38
rs931219953 Chr.4: 99284342-99284342 on Build GRCh38
rs113335478 Chr.4: 99284524-99284524 on Build GRCh38
rs1018663188 Chr.4: 99284446-99284446 on Build GRCh38
rs770538980 Chr.4: 99284466-99284466 on Build GRCh38
rs759629959 Chr.4: 99284382-99284382 on Build GRCh38
rs1049712664 Chr.4: 99284422-99284422 on Build GRCh38
rs574795580 Chr.4: 99284405-99284405 on Build GRCh38
rs774573567 Chr.4: 99284371-99284372 on Build GRCh38
rs750630705 Chr.4: 99284441-99284441 on Build GRCh38
rs767898679 Chr.4: 99284438-99284438 on Build GRCh38
rs201853974 Chr.4: 99284404-99284404 on Build GRCh38
rs763462866 Chr.4: 99284398-99284398 on Build GRCh38
rs868540499 Chr.4: 99284460-99284460 on Build GRCh38
rs778783854 Chr.4: 99284476-99284476 on Build GRCh38
rs144352534 Chr.4: 99284363-99284363 on Build GRCh38
rs147765101 Chr.4: 99284364-99284364 on Build GRCh38
rs752649612 Chr.4: 99284480-99284480 on Build GRCh38
rs777332270 Chr.4: 99284389-99284389 on Build GRCh38
rs780236007 Chr.4: 99284353-99284353 on Build GRCh38
rs376817937 Chr.4: 99284449-99284449 on Build GRCh38
rs774766797 Chr.4: 99284506-99284506 on Build GRCh38
rs753914802 Chr.4: 99284467-99284467 on Build GRCh38
rs776051979 Chr.4: 99284397-99284397 on Build GRCh38
rs747952151 Chr.4: 99284378-99284378 on Build GRCh38
rs769312924 Chr.4: 99284498-99284498 on Build GRCh38
rs996833577 Chr.4: 99284406-99284406 on Build GRCh38

View Full Product Details