Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 232407381-232407614 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALPPL2
Entrez Gene ID
251
Gene Name
alkaline phosphatase, placental like 2
Gene Aliases
ALPG, ALPPL, GCAP
UniGene
Hs.333509
Gene Chromosome Location
Chr.2: 232406842-232410714 on Build GRCh38
Transcripts
NM_031313.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5994136 c.301-2A>G p.? Chr.2: 232407592-232407592 on Build GRCh38
1182624 c.295T>A p.S99T Chr.2: 232407396-232407396 on Build GRCh38
1692163 c.281C>T p.P94L Chr.2: 232407382-232407382 on Build GRCh38
1182625 c.289G>T p.A97S Chr.2: 232407390-232407390 on Build GRCh38
5351968 c.282G>A p.P94P Chr.2: 232407383-232407383 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772838485 Chr.2: 232407575-232407575 on Build GRCh38
rs767747331 Chr.2: 232407440-232407440 on Build GRCh38
rs969083170 Chr.2: 232407530-232407530 on Build GRCh38
rs200330617 Chr.2: 232407408-232407408 on Build GRCh38
rs754524599 Chr.2: 232407544-232407544 on Build GRCh38
rs187153549 Chr.2: 232407429-232407429 on Build GRCh38
rs755682097 Chr.2: 232407444-232407444 on Build GRCh38
rs750176445 Chr.2: 232407422-232407423 on Build GRCh38
rs2945420 Chr.2: 232407436-232407436 on Build GRCh38
rs761616104 Chr.2: 232407402-232407403 on Build GRCh38
rs1043415429 Chr.2: 232407438-232407438 on Build GRCh38
rs1052312061 Chr.2: 232407483-232407483 on Build GRCh38
rs765119037 Chr.2: 232407407-232407420 on Build GRCh38
rs749530689 Chr.2: 232407579-232407579 on Build GRCh38
rs748069109 Chr.2: 232407564-232407564 on Build GRCh38
rs1050479784 Chr.2: 232407406-232407406 on Build GRCh38
rs373835589 Chr.2: 232407428-232407428 on Build GRCh38
rs555647763 Chr.2: 232407560-232407560 on Build GRCh38
rs112182551 Chr.2: 232407584-232407584 on Build GRCh38
rs775395181 Chr.2: 232407416-232407416 on Build GRCh38
rs769844172 Chr.2: 232407588-232407588 on Build GRCh38
rs535702090 Chr.2: 232407460-232407460 on Build GRCh38
rs113818594 Chr.2: 232407533-232407533 on Build GRCh38
rs375205826 Chr.2: 232407589-232407589 on Build GRCh38
rs750112020 Chr.2: 232407442-232407442 on Build GRCh38
rs1005851484 Chr.2: 232407557-232407557 on Build GRCh38
rs759659746 Chr.2: 232407404-232407404 on Build GRCh38
rs896296834 Chr.2: 232407498-232407498 on Build GRCh38
rs771951130 Chr.2: 232407568-232407568 on Build GRCh38
rs112001063 Chr.2: 232407518-232407518 on Build GRCh38
rs746136382 Chr.2: 232407577-232407577 on Build GRCh38
rs1013313158 Chr.2: 232407501-232407501 on Build GRCh38
rs879195192 Chr.2: 232407509-232407509 on Build GRCh38
rs2678517 Chr.2: 232407452-232407452 on Build GRCh38
rs933417965 Chr.2: 232407400-232407400 on Build GRCh38
rs779620249 Chr.2: 232407449-232407449 on Build GRCh38
rs115235444 Chr.2: 232407508-232407508 on Build GRCh38

View Full Product Details