Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 157489045-157489296 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAM19
Entrez Gene ID
8728
Gene Name
ADAM metallopeptidase domain 19
Gene Aliases
FKSG34, MADDAM, MLTNB
UniGene
Hs.483944
Gene Chromosome Location
Chr.5: 157477304-157575823 on Build GRCh38
Transcripts
NM_033274.4 XM_005266003.2 XM_011534682.2 XM_017010009.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4866497 c.2259G>T p.R753S Chr.5: 157489174-157489174 on Build GRCh38
1543086 c.2307G>A p.T769T Chr.5: 157489120-157489120 on Build GRCh38
4983618 c.1487C>G p.P496R Chr.5: 157489139-157489139 on Build GRCh38
5770881 c.2331+5G>C p.? Chr.5: 157489097-157489097 on Build GRCh38
5770882 c.1524+5G>C p.? Chr.5: 157489097-157489097 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751789350 Chr.5: 157489128-157489128 on Build GRCh38
rs753049471 Chr.5: 157489139-157489139 on Build GRCh38
rs969243001 Chr.5: 157489219-157489219 on Build GRCh38
rs914811950 Chr.5: 157489270-157489270 on Build GRCh38
rs1007591734 Chr.5: 157489151-157489151 on Build GRCh38
rs532779242 Chr.5: 157489269-157489269 on Build GRCh38
rs760233391 Chr.5: 157489067-157489067 on Build GRCh38
rs540583846 Chr.5: 157489146-157489146 on Build GRCh38
rs766654040 Chr.5: 157489121-157489121 on Build GRCh38
rs368399855 Chr.5: 157489223-157489223 on Build GRCh38
rs770411254 Chr.5: 157489102-157489104 on Build GRCh38
rs971422780 Chr.5: 157489169-157489169 on Build GRCh38
rs771125721 Chr.5: 157489222-157489222 on Build GRCh38
rs924983399 Chr.5: 157489263-157489263 on Build GRCh38
rs759739166 Chr.5: 157489131-157489131 on Build GRCh38
rs992101242 Chr.5: 157489214-157489214 on Build GRCh38
rs775190280 Chr.5: 157489079-157489079 on Build GRCh38
rs779851864 Chr.5: 157489188-157489188 on Build GRCh38
rs199838986 Chr.5: 157489115-157489115 on Build GRCh38
rs993884128 Chr.5: 157489166-157489166 on Build GRCh38
rs778354767 Chr.5: 157489174-157489174 on Build GRCh38
rs753277652 Chr.5: 157489159-157489159 on Build GRCh38
rs1027557447 Chr.5: 157489133-157489133 on Build GRCh38
rs771895931 Chr.5: 157489078-157489078 on Build GRCh38
rs936451784 Chr.5: 157489268-157489268 on Build GRCh38
rs952076381 Chr.5: 157489156-157489156 on Build GRCh38
rs369947093 Chr.5: 157489096-157489096 on Build GRCh38
rs774660780 Chr.5: 157489065-157489065 on Build GRCh38
rs11466796 Chr.5: 157489272-157489272 on Build GRCh38
rs767890396 Chr.5: 157489137-157489137 on Build GRCh38
rs200701496 Chr.5: 157489194-157489194 on Build GRCh38
rs760640058 Chr.5: 157489087-157489087 on Build GRCh38
rs748159029 Chr.5: 157489202-157489202 on Build GRCh38
rs866194598 Chr.5: 157489112-157489112 on Build GRCh38
rs1024808975 Chr.5: 157489218-157489218 on Build GRCh38
rs763134049 Chr.5: 157489120-157489120 on Build GRCh38
rs768236748 Chr.5: 157489158-157489158 on Build GRCh38
rs756772612 Chr.5: 157489173-157489173 on Build GRCh38
rs776727045 Chr.5: 157489201-157489201 on Build GRCh38
rs963496545 Chr.5: 157489165-157489165 on Build GRCh38
rs759779423 Chr.5: 157489238-157489238 on Build GRCh38
rs149511726 Chr.5: 157489184-157489184 on Build GRCh38
rs745502767 Chr.5: 157489180-157489180 on Build GRCh38
rs201585796 Chr.5: 157489076-157489076 on Build GRCh38

View Full Product Details