Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 51982635-51982908 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACVR1B
Entrez Gene ID
91
Gene Name
activin A receptor type 1B
Gene Aliases
ACTRIB, ACVRLK4, ALK4, SKR2
UniGene
Hs.438918
Gene Chromosome Location
Chr.12: 51951667-51997079 on Build GRCh38
Transcripts
NM_004302.4 NM_020327.3 NM_020328.3 XM_011538966.2 XM_011538967.2 XM_017020199.1 XM_017020200.1 XM_017020201.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5636675 c.840G>A p.W280* Chr.12: 51982714-51982714 on Build GRCh38
4819415 c.917G>T p.R306I Chr.12: 51982791-51982791 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1014651549 Chr.12: 51982745-51982745 on Build GRCh38
rs987647105 Chr.12: 51982705-51982705 on Build GRCh38
rs191020997 Chr.12: 51982739-51982739 on Build GRCh38
rs557422924 Chr.12: 51982710-51982710 on Build GRCh38
rs772140419 Chr.12: 51982772-51982772 on Build GRCh38
rs551375052 Chr.12: 51982733-51982733 on Build GRCh38
rs888601587 Chr.12: 51982727-51982727 on Build GRCh38
rs779924865 Chr.12: 51982734-51982734 on Build GRCh38
rs1029082949 Chr.12: 51982696-51982696 on Build GRCh38
rs1027226983 Chr.12: 51982725-51982725 on Build GRCh38
rs757217593 Chr.12: 51982866-51982866 on Build GRCh38
rs962112771 Chr.12: 51982753-51982753 on Build GRCh38
rs997051385 Chr.12: 51982700-51982700 on Build GRCh38
rs562905372 Chr.12: 51982848-51982848 on Build GRCh38
rs958434031 Chr.12: 51982836-51982836 on Build GRCh38
rs1002507628 Chr.12: 51982770-51982770 on Build GRCh38
rs540762560 Chr.12: 51982682-51982682 on Build GRCh38
rs973642710 Chr.12: 51982853-51982853 on Build GRCh38
rs919564044 Chr.12: 51982856-51982856 on Build GRCh38
rs773517703 Chr.12: 51982806-51982806 on Build GRCh38
rs962248820 Chr.12: 51982834-51982834 on Build GRCh38
rs140512254 Chr.12: 51982830-51982830 on Build GRCh38
rs533142333 Chr.12: 51982702-51982702 on Build GRCh38
rs968515897 Chr.12: 51982875-51982875 on Build GRCh38

View Full Product Details