Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 65778630-65778871 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP5B1
Entrez Gene ID
91056
Gene Name
adaptor related protein complex 5 beta 1 subunit
Gene Aliases
AP-5, PP1030
UniGene
Hs.591957
Gene Chromosome Location
Chr.11: 65773898-65780591 on Build GRCh38
Transcripts
NM_138368.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3810083 c.1835C>T p.A612V Chr.11: 65778658-65778658 on Build GRCh38
6312424 c.1725C>A p.G575G Chr.11: 65778768-65778768 on Build GRCh38
5671581 c.1741C>T p.R581W Chr.11: 65778752-65778752 on Build GRCh38
6430712 c.1720C>T p.Q574* Chr.11: 65778773-65778773 on Build GRCh38
6601803 c.1633G>T p.A545S Chr.11: 65778860-65778860 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770688694 Chr.11: 65778751-65778751 on Build GRCh38
rs745599615 Chr.11: 65778825-65778825 on Build GRCh38
rs373317687 Chr.11: 65778804-65778804 on Build GRCh38
rs914677236 Chr.11: 65778723-65778723 on Build GRCh38
rs763832757 Chr.11: 65778761-65778761 on Build GRCh38
rs749522545 Chr.11: 65778675-65778675 on Build GRCh38
rs775245158 Chr.11: 65778830-65778830 on Build GRCh38
rs774706716 Chr.11: 65778790-65778790 on Build GRCh38
rs775074977 Chr.11: 65778754-65778754 on Build GRCh38
rs374513872 Chr.11: 65778741-65778741 on Build GRCh38
rs375379794 Chr.11: 65778669-65778669 on Build GRCh38
rs749642581 Chr.11: 65778780-65778780 on Build GRCh38
rs763590147 Chr.11: 65778722-65778722 on Build GRCh38
rs752006404 Chr.11: 65778725-65778725 on Build GRCh38
rs746825612 Chr.11: 65778749-65778749 on Build GRCh38
rs758725820 Chr.11: 65778656-65778656 on Build GRCh38
rs111901449 Chr.11: 65778789-65778789 on Build GRCh38
rs762310706 Chr.11: 65778770-65778770 on Build GRCh38
rs146521709 Chr.11: 65778655-65778655 on Build GRCh38
rs896122297 Chr.11: 65778758-65778758 on Build GRCh38
rs1044815492 Chr.11: 65778769-65778769 on Build GRCh38
rs1019532967 Chr.11: 65778713-65778713 on Build GRCh38
rs767874662 Chr.11: 65778704-65778706 on Build GRCh38
rs769973921 Chr.11: 65778668-65778668 on Build GRCh38
rs377608131 Chr.11: 65778752-65778752 on Build GRCh38
rs113072583 Chr.11: 65778660-65778660 on Build GRCh38
rs769804932 Chr.11: 65778828-65778828 on Build GRCh38
rs780167918 Chr.11: 65778740-65778740 on Build GRCh38
rs764768812 Chr.11: 65778711-65778711 on Build GRCh38
rs376122863 Chr.11: 65778760-65778760 on Build GRCh38
rs369808937 Chr.11: 65778736-65778736 on Build GRCh38
rs371620320 Chr.11: 65778659-65778659 on Build GRCh38
rs376592294 Chr.11: 65778805-65778805 on Build GRCh38
rs998940489 Chr.11: 65778827-65778827 on Build GRCh38
rs774214924 Chr.11: 65778851-65778851 on Build GRCh38
rs372682389 Chr.11: 65778701-65778701 on Build GRCh38
rs762683500 Chr.11: 65778717-65778717 on Build GRCh38
rs778189857 Chr.11: 65778657-65778657 on Build GRCh38
rs745969436 Chr.11: 65778667-65778667 on Build GRCh38
rs987495849 Chr.11: 65778687-65778687 on Build GRCh38
rs767990013 Chr.11: 65778773-65778773 on Build GRCh38
rs781777421 Chr.11: 65778738-65778738 on Build GRCh38
rs375914619 Chr.11: 65778702-65778702 on Build GRCh38
rs778961738 Chr.11: 65778748-65778748 on Build GRCh38
rs553712201 Chr.11: 65778678-65778678 on Build GRCh38
rs903232775 Chr.11: 65778813-65778813 on Build GRCh38
rs757106058 Chr.11: 65778808-65778808 on Build GRCh38
rs768924976 Chr.11: 65778747-65778747 on Build GRCh38
rs780948932 Chr.11: 65778822-65778822 on Build GRCh38
rs781751958 Chr.11: 65778662-65778662 on Build GRCh38
rs536395580 Chr.11: 65778768-65778768 on Build GRCh38
rs750825713 Chr.11: 65778736-65778737 on Build GRCh38
rs1048870077 Chr.11: 65778679-65778679 on Build GRCh38
rs752354752 Chr.11: 65778715-65778715 on Build GRCh38
rs754621252 Chr.11: 65778714-65778714 on Build GRCh38
rs769563668 Chr.11: 65778753-65778753 on Build GRCh38
rs768021768 Chr.11: 65778846-65778846 on Build GRCh38
rs748881315 Chr.11: 65778843-65778843 on Build GRCh38
rs369106728 Chr.11: 65778694-65778694 on Build GRCh38
rs370324323 Chr.11: 65778807-65778807 on Build GRCh38
rs760394053 Chr.11: 65778684-65778684 on Build GRCh38
rs750929063 Chr.11: 65778739-65778739 on Build GRCh38
rs770884963 Chr.11: 65778693-65778693 on Build GRCh38
rs932500357 Chr.11: 65778776-65778776 on Build GRCh38

View Full Product Details