Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 100921379-100921648 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP42
Entrez Gene ID
143872
Gene Name
Rho GTPase activating protein 42
Gene Aliases
GRAF3
UniGene
Hs.741465
Gene Chromosome Location
Chr.11: 100687676-100993941 on Build GRCh38
Transcripts
NM_152432.2 XM_011542615.2 XM_011542616.2 XM_011542617.2 XM_017017237.1 XM_017017238.1

SNP Detail

SNP ID Chromosome Location
rs751629132 Chr.11: 100921448-100921448 on Build GRCh38
rs763783297 Chr.11: 100921446-100921447 on Build GRCh38
rs749649644 Chr.11: 100921490-100921490 on Build GRCh38
rs544280034 Chr.11: 100921465-100921465 on Build GRCh38
rs749979796 Chr.11: 100921531-100921531 on Build GRCh38
rs774854009 Chr.11: 100921530-100921530 on Build GRCh38
rs866584638 Chr.11: 100921471-100921471 on Build GRCh38
rs12417981 Chr.11: 100921444-100921444 on Build GRCh38
rs760113834 Chr.11: 100921582-100921582 on Build GRCh38
rs530338405 Chr.11: 100921510-100921510 on Build GRCh38
rs1031603521 Chr.11: 100921603-100921603 on Build GRCh38
rs978189627 Chr.11: 100921596-100921596 on Build GRCh38
rs1012241145 Chr.11: 100921569-100921569 on Build GRCh38
rs1028843223 Chr.11: 100921423-100921423 on Build GRCh38
rs1035000182 Chr.11: 100921507-100921507 on Build GRCh38
rs543699747 Chr.11: 100921563-100921563 on Build GRCh38
rs895131590 Chr.11: 100921512-100921512 on Build GRCh38
rs75746929 Chr.11: 100921604-100921604 on Build GRCh38
rs941169015 Chr.11: 100921426-100921426 on Build GRCh38
rs1015738805 Chr.11: 100921484-100921484 on Build GRCh38
rs968111966 Chr.11: 100921593-100921593 on Build GRCh38
rs961604185 Chr.11: 100921506-100921506 on Build GRCh38
rs750228867 Chr.11: 100921410-100921410 on Build GRCh38
rs752696525 Chr.11: 100921470-100921470 on Build GRCh38
rs1022327643 Chr.11: 100921583-100921583 on Build GRCh38
rs755110400 Chr.11: 100921454-100921454 on Build GRCh38
rs778244723 Chr.11: 100921488-100921488 on Build GRCh38
rs187096137 Chr.11: 100921480-100921480 on Build GRCh38

View Full Product Details