Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 154589280-154589553 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAR
Entrez Gene ID
103
Gene Name
adenosine deaminase, RNA specific
Gene Aliases
ADAR1, AGS6, DRADA, DSH, DSRAD, G1P1, IFI-4, IFI4, K88DSRBP, P136
UniGene
Hs.12341
Gene Chromosome Location
Chr.1: 154582057-154628016 on Build GRCh38
Transcripts
NM_001025107.2 NM_001111.4 NM_001193495.1 NM_015840.3 NM_015841.3 XM_006711109.2 XM_006711111.3 XM_006711112.2 XM_006711113.2 XM_011509060.1 XM_011509061.2 XM_011509062.1 XM_017000037.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3418109 c.2804G>A p.R935H Chr.1: 154589456-154589456 on Build GRCh38
6400740 c.2726G>T p.C909F Chr.1: 154589405-154589405 on Build GRCh38
3418108 c.2675G>A p.R892H Chr.1: 154589456-154589456 on Build GRCh38
4418238 c.2811G>A p.V937V Chr.1: 154589449-154589449 on Build GRCh38
6560937 c.2847C>G p.V949V Chr.1: 154589413-154589413 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754730060 Chr.1: 154589323-154589323 on Build GRCh38
rs768892246 Chr.1: 154589509-154589509 on Build GRCh38
rs965272440 Chr.1: 154589339-154589339 on Build GRCh38
rs780848884 Chr.1: 154589325-154589325 on Build GRCh38
rs751835868 Chr.1: 154589340-154589341 on Build GRCh38
rs376260864 Chr.1: 154589496-154589496 on Build GRCh38
rs760947414 Chr.1: 154589354-154589354 on Build GRCh38
rs746309308 Chr.1: 154589499-154589499 on Build GRCh38
rs35597061 Chr.1: 154589336-154589336 on Build GRCh38
rs201205562 Chr.1: 154589425-154589425 on Build GRCh38
rs142456688 Chr.1: 154589378-154589378 on Build GRCh38
rs982586461 Chr.1: 154589332-154589332 on Build GRCh38
rs201771088 Chr.1: 154589345-154589345 on Build GRCh38
rs766337056 Chr.1: 154589373-154589373 on Build GRCh38
rs777184958 Chr.1: 154589471-154589471 on Build GRCh38
rs377109823 Chr.1: 154589335-154589335 on Build GRCh38
rs368590528 Chr.1: 154589507-154589507 on Build GRCh38
rs1013077618 Chr.1: 154589301-154589301 on Build GRCh38
rs1042509653 Chr.1: 154589466-154589466 on Build GRCh38
rs748858650 Chr.1: 154589488-154589488 on Build GRCh38
rs573632723 Chr.1: 154589308-154589308 on Build GRCh38
rs201315688 Chr.1: 154589362-154589362 on Build GRCh38
rs771435613 Chr.1: 154589343-154589343 on Build GRCh38
rs772630652 Chr.1: 154589506-154589506 on Build GRCh38
rs375147972 Chr.1: 154589344-154589344 on Build GRCh38
rs756670072 Chr.1: 154589495-154589495 on Build GRCh38
rs747744595 Chr.1: 154589330-154589330 on Build GRCh38
rs1037883454 Chr.1: 154589324-154589324 on Build GRCh38
rs753244110 Chr.1: 154589526-154589526 on Build GRCh38
rs958710791 Chr.1: 154589523-154589523 on Build GRCh38
rs775859361 Chr.1: 154589351-154589351 on Build GRCh38
rs879809934 Chr.1: 154589359-154589359 on Build GRCh38
rs372566907 Chr.1: 154589331-154589331 on Build GRCh38
rs144104300 Chr.1: 154589368-154589368 on Build GRCh38
rs755765435 Chr.1: 154589470-154589470 on Build GRCh38
rs766616447 Chr.1: 154589339-154589339 on Build GRCh38
rs754785131 Chr.1: 154589384-154589384 on Build GRCh38
rs536427229 Chr.1: 154589379-154589379 on Build GRCh38
rs371450928 Chr.1: 154589363-154589363 on Build GRCh38
rs906816776 Chr.1: 154589486-154589486 on Build GRCh38
rs1000268386 Chr.1: 154589487-154589487 on Build GRCh38
rs762839847 Chr.1: 154589370-154589370 on Build GRCh38
rs1127309 Chr.1: 154589449-154589449 on Build GRCh38
rs398122892 Chr.1: 154589456-154589456 on Build GRCh38

View Full Product Details