Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 49127881-49128154 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COPS2
Entrez Gene ID
9318
Gene Name
COP9 signalosome subunit 2
Gene Aliases
ALIEN, CSN2, SGN2, TRIP15
UniGene
Hs.369614
Gene Chromosome Location
Chr.15: 49125274-49155657 on Build GRCh38
Transcripts
NM_001143887.1 NM_004236.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5005800 c.1295A>T p.N432I Chr.15: 49127987-49127987 on Build GRCh38
6001648 c.1264A>G p.T422A Chr.15: 49128018-49128018 on Build GRCh38
962460 c.1208A>T p.D403V Chr.15: 49128074-49128074 on Build GRCh38
2216190 c.1259G>A p.R420Q Chr.15: 49128023-49128023 on Build GRCh38
962459 c.1236T>A p.D412E Chr.15: 49128046-49128046 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377673319 Chr.15: 49127946-49127946 on Build GRCh38
rs754590801 Chr.15: 49128090-49128090 on Build GRCh38
rs749289819 Chr.15: 49128118-49128118 on Build GRCh38
rs746513462 Chr.15: 49128069-49128069 on Build GRCh38
rs143666055 Chr.15: 49127982-49127982 on Build GRCh38
rs770987851 Chr.15: 49127989-49127989 on Build GRCh38
rs377183837 Chr.15: 49128023-49128023 on Build GRCh38
rs897982625 Chr.15: 49127940-49127940 on Build GRCh38
rs551528687 Chr.15: 49127928-49127928 on Build GRCh38
rs200813588 Chr.15: 49128019-49128019 on Build GRCh38
rs745320766 Chr.15: 49127914-49127914 on Build GRCh38
rs751003512 Chr.15: 49128046-49128046 on Build GRCh38
rs761517152 Chr.15: 49127947-49127947 on Build GRCh38
rs780737454 Chr.15: 49128091-49128091 on Build GRCh38
rs771754971 Chr.15: 49127919-49127919 on Build GRCh38
rs369542611 Chr.15: 49128013-49128013 on Build GRCh38
rs769626565 Chr.15: 49128109-49128109 on Build GRCh38
rs182165965 Chr.15: 49128110-49128110 on Build GRCh38
rs1036131602 Chr.15: 49128108-49128108 on Build GRCh38
rs772129800 Chr.15: 49128128-49128128 on Build GRCh38
rs765813477 Chr.15: 49128036-49128036 on Build GRCh38
rs779767814 Chr.15: 49128061-49128061 on Build GRCh38
rs769677428 Chr.15: 49127966-49127966 on Build GRCh38
rs757582940 Chr.15: 49128025-49128025 on Build GRCh38
rs370391204 Chr.15: 49128054-49128054 on Build GRCh38
rs1009037344 Chr.15: 49127909-49127909 on Build GRCh38
rs767625268 Chr.15: 49127987-49127987 on Build GRCh38
rs201832558 Chr.15: 49128042-49128042 on Build GRCh38
rs773157842 Chr.15: 49127978-49127978 on Build GRCh38
rs746739053 Chr.15: 49127938-49127938 on Build GRCh38
rs751293209 Chr.15: 49127932-49127938 on Build GRCh38
rs747929203 Chr.15: 49128093-49128093 on Build GRCh38
rs768322431 Chr.15: 49127945-49127945 on Build GRCh38
rs754794557 Chr.15: 49128121-49128122 on Build GRCh38

View Full Product Details