Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ASAP1
Entrez Gene ID
50807
Gene Name
ArfGAP with SH3 domain, ankyrin repeat and PH domain 1
Gene Aliases
AMAP1, CENTB4, DDEF1, PAG2, PAP, ZG14P
Chromosome Location
Chr.8: 130052104 - 130443848 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001247996.1 NP_001234925.1 18 - 19 1848 111
NM_018482.3 NP_060952.2 17 - 18 1746 111
XM_005250925.1 XP_005250982.1 18 - 19 1755 111
XM_006716563.3 XP_006716626.1 18 - 19 1574 111
XM_006716564.1 XP_006716627.1 19 - 20 1733 111
XM_006716565.3 XP_006716628.1 17 - 18 1570 111
XM_006716566.1 XP_006716629.1 18 - 19 1755 111
XM_011517052.2 XP_011515354.1 18 - 19 1571 111
XM_011517053.1 XP_011515355.1 18 - 19 1777 111
XM_017013467.1 XP_016868956.1 16 - 17 1592 111
XM_017013468.1 XP_016868957.1 17 - 18 1816 111
XM_017013469.1 XP_016868958.1 17 - 18 1746 111
GenBank mRNA AB033075.1 - 11 - 12 979 111
BC137135.1 - 16 - 17 1543 111
BC144493.1 - 17 - 18 1645 111
BX537768.1 - 10 - 11 999 111

View Full Product Details