Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 32545200-32545473 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKAP6
Entrez Gene ID
9472
Gene Name
A-kinase anchoring protein 6
Gene Aliases
ADAP100, ADAP6, AKAP100, PRKA6, mAKAP
UniGene
Hs.509083
Gene Chromosome Location
Chr.14: 32329273-32833999 on Build GRCh38
Transcripts
NM_004274.4 XM_017021808.1 XM_017021809.1 XM_017021810.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5519248 c.720A>G p.L240L Chr.14: 32545373-32545373 on Build GRCh38
5032700 c.581G>A p.R194Q Chr.14: 32545234-32545234 on Build GRCh38
1677927 c.676G>A p.E226K Chr.14: 32545329-32545329 on Build GRCh38
1244746 c.612A>G p.S204S Chr.14: 32545265-32545265 on Build GRCh38
4558793 c.780G>A p.E260E Chr.14: 32545433-32545433 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747787990 Chr.14: 32545265-32545265 on Build GRCh38
rs776194183 Chr.14: 32545255-32545255 on Build GRCh38
rs139779310 Chr.14: 32545296-32545296 on Build GRCh38
rs780381859 Chr.14: 32545245-32545245 on Build GRCh38
rs779406344 Chr.14: 32545234-32545234 on Build GRCh38
rs754081380 Chr.14: 32545232-32545232 on Build GRCh38
rs761996053 Chr.14: 32545440-32545440 on Build GRCh38
rs61757554 Chr.14: 32545250-32545250 on Build GRCh38
rs754028032 Chr.14: 32545431-32545431 on Build GRCh38
rs890176378 Chr.14: 32545260-32545260 on Build GRCh38
rs145116316 Chr.14: 32545368-32545368 on Build GRCh38
rs929313640 Chr.14: 32545320-32545320 on Build GRCh38
rs916435002 Chr.14: 32545445-32545445 on Build GRCh38
rs776913909 Chr.14: 32545340-32545340 on Build GRCh38
rs759587062 Chr.14: 32545406-32545406 on Build GRCh38
rs1054855838 Chr.14: 32545377-32545377 on Build GRCh38
rs868161302 Chr.14: 32545329-32545329 on Build GRCh38
rs201721425 Chr.14: 32545293-32545293 on Build GRCh38
rs1000136984 Chr.14: 32545388-32545388 on Build GRCh38
rs764264334 Chr.14: 32545409-32545409 on Build GRCh38
rs546051639 Chr.14: 32545305-32545305 on Build GRCh38
rs757619323 Chr.14: 32545233-32545233 on Build GRCh38
rs1032583521 Chr.14: 32545403-32545403 on Build GRCh38
rs768128356 Chr.14: 32545253-32545253 on Build GRCh38
rs746159969 Chr.14: 32545239-32545239 on Build GRCh38
rs931635354 Chr.14: 32545235-32545235 on Build GRCh38
rs758792601 Chr.14: 32545243-32545243 on Build GRCh38
rs33936626 Chr.14: 32545343-32545343 on Build GRCh38
rs149076778 Chr.14: 32545372-32545372 on Build GRCh38
rs976176666 Chr.14: 32545264-32545264 on Build GRCh38
rs983452601 Chr.14: 32545328-32545328 on Build GRCh38

View Full Product Details