Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 15393930-15394179 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGMO
Entrez Gene ID
392636
Gene Name
alkylglycerol monooxygenase
Gene Aliases
TMEM195
UniGene
Hs.670634
Gene Chromosome Location
Chr.7: 15117223-15562015 on Build GRCh38
Transcripts
NM_001004320.1 XM_006715730.1 XM_006715731.3 XM_011515402.2 XM_017012204.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4155666 c.664A>T p.R222W Chr.7: 15394125-15394125 on Build GRCh38
5887965 c.653C>A p.P218H Chr.7: 15394136-15394136 on Build GRCh38
5872330 c.621_622CC>TT p.L208F Chr.7: 15394167-15394168 on Build GRCh38
485110 c.636A>G p.E212E Chr.7: 15394153-15394153 on Build GRCh38
3358112 c.656G>A p.S219N Chr.7: 15394133-15394133 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs148081590 Chr.7: 15394042-15394042 on Build GRCh38
rs923555972 Chr.7: 15394133-15394134 on Build GRCh38
rs999465663 Chr.7: 15394049-15394049 on Build GRCh38
rs755989727 Chr.7: 15394124-15394124 on Build GRCh38
rs1040577987 Chr.7: 15393991-15393994 on Build GRCh38
rs772636718 Chr.7: 15394084-15394085 on Build GRCh38
rs748678920 Chr.7: 15394080-15394082 on Build GRCh38
rs918070424 Chr.7: 15393956-15393956 on Build GRCh38
rs201080212 Chr.7: 15394106-15394106 on Build GRCh38
rs750140675 Chr.7: 15394123-15394123 on Build GRCh38
rs913879210 Chr.7: 15394139-15394139 on Build GRCh38
rs893260383 Chr.7: 15394022-15394022 on Build GRCh38
rs763654051 Chr.7: 15394115-15394115 on Build GRCh38
rs747514990 Chr.7: 15394098-15394098 on Build GRCh38
rs754063976 Chr.7: 15394085-15394085 on Build GRCh38
rs771290913 Chr.7: 15394104-15394104 on Build GRCh38
rs374622798 Chr.7: 15394151-15394151 on Build GRCh38
rs549438301 Chr.7: 15394063-15394063 on Build GRCh38
rs890961146 Chr.7: 15394029-15394029 on Build GRCh38
rs142034204 Chr.7: 15394152-15394152 on Build GRCh38
rs1045290236 Chr.7: 15393965-15393965 on Build GRCh38
rs372277184 Chr.7: 15394062-15394062 on Build GRCh38
rs751519665 Chr.7: 15394073-15394073 on Build GRCh38
rs150962030 Chr.7: 15394136-15394136 on Build GRCh38
rs940958075 Chr.7: 15393989-15393989 on Build GRCh38
rs531224100 Chr.7: 15394060-15394060 on Build GRCh38
rs995158377 Chr.7: 15394056-15394056 on Build GRCh38
rs566081795 Chr.7: 15393998-15393998 on Build GRCh38
rs908013521 Chr.7: 15393985-15393985 on Build GRCh38
rs560030506 Chr.7: 15393997-15393997 on Build GRCh38
rs778408810 Chr.7: 15394095-15394095 on Build GRCh38
rs878886947 Chr.7: 15394034-15394034 on Build GRCh38
rs1010499442 Chr.7: 15394014-15394014 on Build GRCh38
rs749508611 Chr.7: 15394086-15394086 on Build GRCh38
rs762461245 Chr.7: 15394067-15394067 on Build GRCh38
rs762448591 Chr.7: 15394114-15394114 on Build GRCh38
rs200156433 Chr.7: 15394132-15394132 on Build GRCh38
rs948328514 Chr.7: 15393958-15393958 on Build GRCh38
rs779303970 Chr.7: 15394088-15394088 on Build GRCh38
rs370101645 Chr.7: 15394118-15394118 on Build GRCh38
rs747034817 Chr.7: 15394087-15394088 on Build GRCh38
rs746265175 Chr.7: 15394107-15394107 on Build GRCh38
rs201537987 Chr.7: 15394100-15394101 on Build GRCh38
rs748183815 Chr.7: 15394090-15394090 on Build GRCh38
rs541624918 Chr.7: 15393976-15393976 on Build GRCh38
rs767526135 Chr.7: 15394122-15394122 on Build GRCh38
rs147049004 Chr.7: 15394039-15394039 on Build GRCh38
rs757063669 Chr.7: 15394075-15394075 on Build GRCh38
rs1049260983 Chr.7: 15393979-15393979 on Build GRCh38
rs767265165 Chr.7: 15394079-15394079 on Build GRCh38
rs1011628842 Chr.7: 15394031-15394031 on Build GRCh38
rs1050944592 Chr.7: 15394016-15394016 on Build GRCh38
rs761697067 Chr.7: 15394120-15394120 on Build GRCh38
rs776820668 Chr.7: 15394105-15394105 on Build GRCh38
rs775125939 Chr.7: 15394110-15394110 on Build GRCh38
rs769377948 Chr.7: 15394109-15394109 on Build GRCh38
rs892528899 Chr.7: 15394009-15394009 on Build GRCh38
rs143437900 Chr.7: 15394135-15394135 on Build GRCh38
rs773882530 Chr.7: 15394085-15394088 on Build GRCh38
rs185614914 Chr.7: 15393974-15393974 on Build GRCh38
rs201523312 Chr.7: 15394092-15394092 on Build GRCh38
rs367702435 Chr.7: 15394071-15394071 on Build GRCh38

View Full Product Details