Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 72911725-72911996 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALDH1A1
Entrez Gene ID
216
Gene Name
aldehyde dehydrogenase 1 family member A1
Gene Aliases
ALDC, ALDH-E1, ALDH1, ALDH11, HEL-9, HEL-S-53e, HEL12, PUMB1, RALDH1
UniGene
Hs.76392
Gene Chromosome Location
Chr.9: 72900662-72953317 on Build GRCh38
Transcripts
NM_000689.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6598174 c.1179G>A p.E393E Chr.9: 72911979-72911979 on Build GRCh38
1109994 c.1184G>A p.R395H Chr.9: 72911974-72911974 on Build GRCh38
3907924 c.1183C>T p.R395C Chr.9: 72911975-72911975 on Build GRCh38
6183467 c.1177G>A p.E393K Chr.9: 72911981-72911981 on Build GRCh38
7152729 c.1195G>C p.E399Q Chr.9: 72911963-72911963 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1046887251 Chr.9: 72911748-72911748 on Build GRCh38
rs956718285 Chr.9: 72911905-72911905 on Build GRCh38
rs747862603 Chr.9: 72911919-72911919 on Build GRCh38
rs772537628 Chr.9: 72911944-72911944 on Build GRCh38
rs938378164 Chr.9: 72911814-72911814 on Build GRCh38
rs764465241 Chr.9: 72911961-72911961 on Build GRCh38
rs1028982069 Chr.9: 72911948-72911948 on Build GRCh38
rs149002760 Chr.9: 72911826-72911826 on Build GRCh38
rs1000902207 Chr.9: 72911889-72911889 on Build GRCh38
rs1053355736 Chr.9: 72911781-72911781 on Build GRCh38
rs373784862 Chr.9: 72911930-72911930 on Build GRCh38
rs751612395 Chr.9: 72911910-72911910 on Build GRCh38
rs894832422 Chr.9: 72911807-72911807 on Build GRCh38
rs1014759888 Chr.9: 72911883-72911883 on Build GRCh38
rs878858808 Chr.9: 72911957-72911957 on Build GRCh38
rs746424880 Chr.9: 72911942-72911942 on Build GRCh38
rs375889705 Chr.9: 72911949-72911949 on Build GRCh38
rs886768573 Chr.9: 72911757-72911757 on Build GRCh38
rs896746559 Chr.9: 72911830-72911830 on Build GRCh38
rs754950709 Chr.9: 72911913-72911913 on Build GRCh38
rs749467487 Chr.9: 72911935-72911935 on Build GRCh38
rs199619658 Chr.9: 72911927-72911927 on Build GRCh38
rs113786983 Chr.9: 72911774-72911774 on Build GRCh38
rs755899140 Chr.9: 72911920-72911920 on Build GRCh38
rs568524808 Chr.9: 72911875-72911875 on Build GRCh38
rs372316685 Chr.9: 72911902-72911902 on Build GRCh38
rs770918243 Chr.9: 72911937-72911937 on Build GRCh38
rs145863014 Chr.9: 72911845-72911845 on Build GRCh38
rs8187974 Chr.9: 72911950-72911950 on Build GRCh38
rs1012385598 Chr.9: 72911844-72911844 on Build GRCh38
rs774402809 Chr.9: 72911941-72911941 on Build GRCh38
rs903907244 Chr.9: 72911872-72911872 on Build GRCh38
rs781178924 Chr.9: 72911914-72911914 on Build GRCh38
rs752185042 Chr.9: 72911908-72911911 on Build GRCh38
rs373206952 Chr.9: 72911952-72911952 on Build GRCh38

View Full Product Details