Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 71738773-71739016 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP1G1
Entrez Gene ID
164
Gene Name
adaptor related protein complex 1 gamma 1 subunit
Gene Aliases
ADTG, CLAPG1
UniGene
Hs.461253
Gene Chromosome Location
Chr.16: 71729000-71809073 on Build GRCh38
Transcripts
NM_001030007.1 NM_001128.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6285379 c.2211C>A p.A737A Chr.16: 71738999-71738999 on Build GRCh38
6206198 c.2336A>T p.K779M Chr.16: 71738943-71738943 on Build GRCh38
6285378 c.2220C>A p.A740A Chr.16: 71738999-71738999 on Build GRCh38
4414218 c.2218A>T p.S740C Chr.16: 71738992-71738992 on Build GRCh38
4414217 c.2227A>T p.S743C Chr.16: 71738992-71738992 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs946368074 Chr.16: 71738854-71738854 on Build GRCh38
rs9929582 Chr.16: 71738881-71738881 on Build GRCh38
rs547908245 Chr.16: 71738864-71738864 on Build GRCh38
rs371660834 Chr.16: 71738892-71738892 on Build GRCh38
rs751111867 Chr.16: 71738924-71738924 on Build GRCh38
rs760012198 Chr.16: 71738893-71738893 on Build GRCh38
rs371126580 Chr.16: 71738809-71738811 on Build GRCh38
rs1017141527 Chr.16: 71738938-71738938 on Build GRCh38
rs756700572 Chr.16: 71738928-71738928 on Build GRCh38
rs1003726626 Chr.16: 71738884-71738884 on Build GRCh38
rs879076046 Chr.16: 71738881-71738882 on Build GRCh38
rs745506245 Chr.16: 71738954-71738954 on Build GRCh38
rs149867815 Chr.16: 71738901-71738901 on Build GRCh38
rs904764 Chr.16: 71738892-71738892 on Build GRCh38
rs377472148 Chr.16: 71738910-71738910 on Build GRCh38
rs960713774 Chr.16: 71738835-71738835 on Build GRCh38
rs199808145 Chr.16: 71738921-71738921 on Build GRCh38
rs935130568 Chr.16: 71738813-71738813 on Build GRCh38
rs543879878 Chr.16: 71738903-71738903 on Build GRCh38
rs915128527 Chr.16: 71738825-71738825 on Build GRCh38
rs747615231 Chr.16: 71738924-71738926 on Build GRCh38
rs568896446 Chr.16: 71738812-71738812 on Build GRCh38
rs1030944600 Chr.16: 71738830-71738830 on Build GRCh38
rs1013595855 Chr.16: 71738840-71738840 on Build GRCh38
rs778151169 Chr.16: 71738860-71738860 on Build GRCh38
rs67141904 Chr.16: 71738881-71738881 on Build GRCh38
rs780956795 Chr.16: 71738945-71738945 on Build GRCh38
rs755945949 Chr.16: 71738962-71738962 on Build GRCh38
rs779658464 Chr.16: 71738972-71738972 on Build GRCh38
rs112238313 Chr.16: 71738821-71738821 on Build GRCh38
rs62053157 Chr.16: 71738894-71738894 on Build GRCh38
rs537695512 Chr.16: 71738981-71738981 on Build GRCh38
rs753214058 Chr.16: 71738909-71738909 on Build GRCh38

View Full Product Details