Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 47667496-47667767 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGBL2
Entrez Gene ID
79841
Gene Name
ATP/GTP binding protein like 2
Gene Aliases
CCP2
UniGene
Hs.147377
Gene Chromosome Location
Chr.11: 47659168-47715397 on Build GRCh38
Transcripts
NM_024783.3 XM_005253138.4 XM_005253139.4 XM_005253140.4 XM_006718327.3 XM_006718328.3 XM_011520372.2 XM_011520373.2 XM_011520374.2 XM_011520375.2 XM_011520376.2 XM_011520377.2 XM_011520378.2 XM_011520379.2 XM_011520381.2 XM_017018331.1 XM_017018332.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7153102 c.2223G>A p.Q741Q Chr.11: 47667688-47667688 on Build GRCh38
4192058 c.2304T>A p.N768K Chr.11: 47667607-47667607 on Build GRCh38
4033235 c.2302A>C p.N768H Chr.11: 47667609-47667609 on Build GRCh38
3447856 c.2229G>A p.K743K Chr.11: 47667682-47667682 on Build GRCh38
1746277 c.2227A>T p.K743* Chr.11: 47667684-47667684 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776937327 Chr.11: 47667542-47667542 on Build GRCh38
rs748845312 Chr.11: 47667676-47667676 on Build GRCh38
rs377578482 Chr.11: 47667699-47667699 on Build GRCh38
rs202194321 Chr.11: 47667634-47667634 on Build GRCh38
rs772198482 Chr.11: 47667688-47667688 on Build GRCh38
rs762516134 Chr.11: 47667582-47667582 on Build GRCh38
rs748480950 Chr.11: 47667543-47667543 on Build GRCh38
rs772854330 Chr.11: 47667672-47667672 on Build GRCh38
rs955360361 Chr.11: 47667732-47667732 on Build GRCh38
rs769083984 Chr.11: 47667541-47667541 on Build GRCh38
rs773358255 Chr.11: 47667607-47667608 on Build GRCh38
rs369826804 Chr.11: 47667744-47667744 on Build GRCh38
rs779740246 Chr.11: 47667729-47667729 on Build GRCh38
rs370219080 Chr.11: 47667564-47667564 on Build GRCh38
rs751638690 Chr.11: 47667730-47667730 on Build GRCh38
rs747577453 Chr.11: 47667661-47667661 on Build GRCh38
rs539989578 Chr.11: 47667608-47667608 on Build GRCh38
rs755474349 Chr.11: 47667662-47667662 on Build GRCh38
rs758407250 Chr.11: 47667521-47667521 on Build GRCh38
rs767458914 Chr.11: 47667617-47667617 on Build GRCh38
rs765750786 Chr.11: 47667602-47667602 on Build GRCh38
rs766447259 Chr.11: 47667634-47667635 on Build GRCh38
rs780613193 Chr.11: 47667530-47667530 on Build GRCh38
rs201481571 Chr.11: 47667682-47667682 on Build GRCh38
rs576901846 Chr.11: 47667560-47667560 on Build GRCh38
rs373392891 Chr.11: 47667568-47667568 on Build GRCh38
rs977275474 Chr.11: 47667741-47667741 on Build GRCh38
rs755965090 Chr.11: 47667632-47667632 on Build GRCh38
rs776595317 Chr.11: 47667658-47667659 on Build GRCh38
rs1029588162 Chr.11: 47667718-47667718 on Build GRCh38
rs760554344 Chr.11: 47667708-47667708 on Build GRCh38
rs377064786 Chr.11: 47667633-47667633 on Build GRCh38
rs765108242 Chr.11: 47667678-47667680 on Build GRCh38
rs763920978 Chr.11: 47667710-47667710 on Build GRCh38
rs141431736 Chr.11: 47667693-47667693 on Build GRCh38
rs368783386 Chr.11: 47667681-47667681 on Build GRCh38
rs747414679 Chr.11: 47667537-47667537 on Build GRCh38
rs192558333 Chr.11: 47667670-47667670 on Build GRCh38
rs770136980 Chr.11: 47667671-47667671 on Build GRCh38
rs760684575 Chr.11: 47667608-47667608 on Build GRCh38
rs769506962 Chr.11: 47667667-47667667 on Build GRCh38
rs752592989 Chr.11: 47667622-47667622 on Build GRCh38
rs774370452 Chr.11: 47667606-47667606 on Build GRCh38
rs996854782 Chr.11: 47667692-47667692 on Build GRCh38
rs759065796 Chr.11: 47667684-47667684 on Build GRCh38
rs766588190 Chr.11: 47667723-47667723 on Build GRCh38
rs750053721 Chr.11: 47667636-47667636 on Build GRCh38
rs570874643 Chr.11: 47667722-47667722 on Build GRCh38

View Full Product Details