Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 38389739-38390010 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMPH
Entrez Gene ID
273
Gene Name
amphiphysin
Gene Aliases
AMPH1
UniGene
Hs.592182
Gene Chromosome Location
Chr.7: 38383694-38631567 on Build GRCh38
Transcripts
NM_001635.3 NM_139316.2 XM_006715689.3 XM_006715690.3 XM_011515271.2 XM_017011995.1 XM_017011996.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5038995 c.1975G>T p.D659Y Chr.7: 38389809-38389809 on Build GRCh38
4947711 c.1791T>C p.D597D Chr.7: 38389867-38389867 on Build GRCh38
7249845 c.1753G>T p.V585L Chr.7: 38389905-38389905 on Build GRCh38
1089472 c.1919A>G p.E640G Chr.7: 38389865-38389865 on Build GRCh38
351611 c.1975G>C p.D659H Chr.7: 38389809-38389809 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs878981509 Chr.7: 38389958-38389958 on Build GRCh38
rs200109991 Chr.7: 38389866-38389866 on Build GRCh38
rs370149433 Chr.7: 38389919-38389919 on Build GRCh38
rs373411830 Chr.7: 38389770-38389770 on Build GRCh38
rs753997975 Chr.7: 38389925-38389925 on Build GRCh38
rs377538910 Chr.7: 38389881-38389881 on Build GRCh38
rs867571728 Chr.7: 38389847-38389847 on Build GRCh38
rs750837865 Chr.7: 38389901-38389901 on Build GRCh38
rs932867620 Chr.7: 38389777-38389777 on Build GRCh38
rs35606872 Chr.7: 38389972-38389972 on Build GRCh38
rs36122520 Chr.7: 38389951-38389951 on Build GRCh38
rs766643117 Chr.7: 38389916-38389916 on Build GRCh38
rs755050238 Chr.7: 38389938-38389938 on Build GRCh38
rs879118313 Chr.7: 38389930-38389930 on Build GRCh38
rs770659075 Chr.7: 38389819-38389819 on Build GRCh38
rs759140530 Chr.7: 38389828-38389828 on Build GRCh38
rs200440033 Chr.7: 38389943-38389943 on Build GRCh38
rs765991040 Chr.7: 38389800-38389800 on Build GRCh38
rs748016969 Chr.7: 38389789-38389789 on Build GRCh38
rs775135384 Chr.7: 38389859-38389859 on Build GRCh38
rs6977263 Chr.7: 38389769-38389769 on Build GRCh38
rs34824107 Chr.7: 38389983-38389983 on Build GRCh38
rs762466435 Chr.7: 38389878-38389878 on Build GRCh38
rs746760641 Chr.7: 38389806-38389806 on Build GRCh38
rs771996761 Chr.7: 38389798-38389798 on Build GRCh38
rs554921009 Chr.7: 38389768-38389768 on Build GRCh38
rs964117064 Chr.7: 38389887-38389887 on Build GRCh38
rs997362813 Chr.7: 38389914-38389914 on Build GRCh38
rs776108511 Chr.7: 38389826-38389826 on Build GRCh38
rs760905601 Chr.7: 38389911-38389911 on Build GRCh38
rs1050019520 Chr.7: 38389912-38389912 on Build GRCh38
rs564091142 Chr.7: 38389920-38389920 on Build GRCh38

View Full Product Details