Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 223758604-223758877 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP1S3
Entrez Gene ID
130340
Gene Name
adaptor related protein complex 1 sigma 3 subunit
Gene Aliases
PSORS15
UniGene
Hs.632555
Gene Chromosome Location
Chr.2: 223755330-223837602 on Build GRCh38
Transcripts
NM_001039569.1 NR_110905.1 NR_110906.1 XM_011510600.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1017069 c.439G>A p.E147K Chr.2: 223758741-223758741 on Build GRCh38
5679628 c.431C>A p.T144K Chr.2: 223758749-223758749 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs922820988 Chr.2: 223758679-223758679 on Build GRCh38
rs941439858 Chr.2: 223758738-223758738 on Build GRCh38
rs757952789 Chr.2: 223758761-223758761 on Build GRCh38
rs371749232 Chr.2: 223758692-223758692 on Build GRCh38
rs757132831 Chr.2: 223758785-223758785 on Build GRCh38
rs557525015 Chr.2: 223758685-223758685 on Build GRCh38
rs761047979 Chr.2: 223758735-223758735 on Build GRCh38
rs139347744 Chr.2: 223758662-223758662 on Build GRCh38
rs746842982 Chr.2: 223758677-223758677 on Build GRCh38
rs780974940 Chr.2: 223758787-223758787 on Build GRCh38
rs372622960 Chr.2: 223758691-223758691 on Build GRCh38
rs754301459 Chr.2: 223758748-223758748 on Build GRCh38
rs769818919 Chr.2: 223758802-223758802 on Build GRCh38
rs151312965 Chr.2: 223758658-223758658 on Build GRCh38
rs747611108 Chr.2: 223758670-223758670 on Build GRCh38
rs746612168 Chr.2: 223758784-223758784 on Build GRCh38
rs767764455 Chr.2: 223758727-223758727 on Build GRCh38
rs537681063 Chr.2: 223758673-223758673 on Build GRCh38
rs752303998 Chr.2: 223758759-223758759 on Build GRCh38
rs917835051 Chr.2: 223758733-223758733 on Build GRCh38
rs184644015 Chr.2: 223758758-223758758 on Build GRCh38
rs768622808 Chr.2: 223758694-223758694 on Build GRCh38
rs750601010 Chr.2: 223758728-223758728 on Build GRCh38
rs376934833 Chr.2: 223758647-223758647 on Build GRCh38
rs745636981 Chr.2: 223758790-223758790 on Build GRCh38
rs774547757 Chr.2: 223758704-223758704 on Build GRCh38
rs757738145 Chr.2: 223758798-223758798 on Build GRCh38
rs951632228 Chr.2: 223758688-223758688 on Build GRCh38
rs1011108460 Chr.2: 223758637-223758637 on Build GRCh38
rs982966220 Chr.2: 223758712-223758712 on Build GRCh38
rs749600722 Chr.2: 223758797-223758797 on Build GRCh38
rs142453419 Chr.2: 223758657-223758657 on Build GRCh38
rs374882581 Chr.2: 223758713-223758713 on Build GRCh38
rs190326614 Chr.2: 223758803-223758803 on Build GRCh38
rs925071822 Chr.2: 223758770-223758770 on Build GRCh38
rs777602392 Chr.2: 223758782-223758782 on Build GRCh38
rs1007050093 Chr.2: 223758646-223758646 on Build GRCh38

View Full Product Details