Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 87531311-87531566 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCB1
Entrez Gene ID
5243
Gene Name
ATP binding cassette subfamily B member 1
Gene Aliases
ABC20, CD243, CLCS, GP170, MDR1, P-GP, PGY1
UniGene
Hs.489033
Gene Chromosome Location
Chr.7: 87503863-87713323 on Build GRCh38
Transcripts
NM_000927.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5439010 c.2511C>A p.T837T Chr.7: 87531468-87531468 on Build GRCh38
1568835 c.2659A>G p.K887E Chr.7: 87531320-87531320 on Build GRCh38
5873289 c.2626A>G p.M876V Chr.7: 87531353-87531353 on Build GRCh38
1243950 c.2528T>G p.L843R Chr.7: 87531451-87531451 on Build GRCh38
3883054 c.2665G>T p.E889* Chr.7: 87531314-87531314 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs28381966 Chr.7: 87531474-87531474 on Build GRCh38
rs199599822 Chr.7: 87531499-87531499 on Build GRCh38
rs36105130 Chr.7: 87531432-87531432 on Build GRCh38
rs762672319 Chr.7: 87531533-87531533 on Build GRCh38
rs775305519 Chr.7: 87531502-87531502 on Build GRCh38
rs200203709 Chr.7: 87531344-87531344 on Build GRCh38
rs752216494 Chr.7: 87531500-87531500 on Build GRCh38
rs200711678 Chr.7: 87531527-87531527 on Build GRCh38
rs778283893 Chr.7: 87531383-87531383 on Build GRCh38
rs746152564 Chr.7: 87531415-87531415 on Build GRCh38
rs772374336 Chr.7: 87531420-87531420 on Build GRCh38
rs28381967 Chr.7: 87531473-87531473 on Build GRCh38
rs557586335 Chr.7: 87531422-87531422 on Build GRCh38
rs767089531 Chr.7: 87531490-87531490 on Build GRCh38
rs773175021 Chr.7: 87531442-87531442 on Build GRCh38
rs199661708 Chr.7: 87531411-87531411 on Build GRCh38
rs773262840 Chr.7: 87531463-87531463 on Build GRCh38
rs780701513 Chr.7: 87531339-87531339 on Build GRCh38
rs367909991 Chr.7: 87531348-87531348 on Build GRCh38
rs374182865 Chr.7: 87531429-87531429 on Build GRCh38
rs371274259 Chr.7: 87531421-87531421 on Build GRCh38
rs747022752 Chr.7: 87531423-87531423 on Build GRCh38
rs201741691 Chr.7: 87531399-87531399 on Build GRCh38
rs200496828 Chr.7: 87531472-87531472 on Build GRCh38
rs762765487 Chr.7: 87531470-87531470 on Build GRCh38
rs749745819 Chr.7: 87531388-87531388 on Build GRCh38
rs565619660 Chr.7: 87531358-87531358 on Build GRCh38
rs1000911090 Chr.7: 87531397-87531397 on Build GRCh38
rs539556127 Chr.7: 87531389-87531389 on Build GRCh38
rs2032581 Chr.7: 87531494-87531494 on Build GRCh38
rs755679093 Chr.7: 87531516-87531516 on Build GRCh38
rs754409465 Chr.7: 87531528-87531528 on Build GRCh38
rs368935884 Chr.7: 87531518-87531518 on Build GRCh38

View Full Product Details