Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 21250245-21250476 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKS4B
Entrez Gene ID
257629
Gene Name
ankyrin repeat and sterile alpha motif domain containing 4B
Gene Aliases
HARP
UniGene
Hs.115959
Gene Chromosome Location
Chr.16: 21233695-21252429 on Build GRCh38
Transcripts
NM_145865.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3507440 c.728C>T p.S243L Chr.16: 21250294-21250294 on Build GRCh38
6578243 c.763G>C p.E255Q Chr.16: 21250329-21250329 on Build GRCh38
968425 c.687C>T p.N229N Chr.16: 21250253-21250253 on Build GRCh38
3507441 c.815G>A p.G272E Chr.16: 21250381-21250381 on Build GRCh38
968426 c.856G>T p.D286Y Chr.16: 21250422-21250422 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749814184 Chr.16: 21250289-21250289 on Build GRCh38
rs769678627 Chr.16: 21250437-21250437 on Build GRCh38
rs927033805 Chr.16: 21250409-21250409 on Build GRCh38
rs980284367 Chr.16: 21250395-21250395 on Build GRCh38
rs779447867 Chr.16: 21250394-21250394 on Build GRCh38
rs950785962 Chr.16: 21250332-21250332 on Build GRCh38
rs756297978 Chr.16: 21250366-21250366 on Build GRCh38
rs768357197 Chr.16: 21250319-21250319 on Build GRCh38
rs748003891 Chr.16: 21250434-21250434 on Build GRCh38
rs17707647 Chr.16: 21250364-21250364 on Build GRCh38
rs746345226 Chr.16: 21250400-21250400 on Build GRCh38
rs746656156 Chr.16: 21250413-21250413 on Build GRCh38
rs548769339 Chr.16: 21250425-21250425 on Build GRCh38
rs760333449 Chr.16: 21250313-21250313 on Build GRCh38
rs779569210 Chr.16: 21250412-21250412 on Build GRCh38
rs765325045 Chr.16: 21250338-21250338 on Build GRCh38
rs776429674 Chr.16: 21250322-21250322 on Build GRCh38
rs994813981 Chr.16: 21250304-21250304 on Build GRCh38
rs369712734 Chr.16: 21250324-21250324 on Build GRCh38
rs988128401 Chr.16: 21250444-21250444 on Build GRCh38
rs999253418 Chr.16: 21250301-21250301 on Build GRCh38
rs778154762 Chr.16: 21250288-21250288 on Build GRCh38
rs759727437 Chr.16: 21250347-21250347 on Build GRCh38
rs375261621 Chr.16: 21250367-21250367 on Build GRCh38
rs757879437 Chr.16: 21250411-21250411 on Build GRCh38
rs754271798 Chr.16: 21250368-21250368 on Build GRCh38
rs771545401 Chr.16: 21250300-21250300 on Build GRCh38
rs757608549 Chr.16: 21250369-21250369 on Build GRCh38
rs767591528 Chr.16: 21250351-21250351 on Build GRCh38
rs750362559 Chr.16: 21250346-21250346 on Build GRCh38
rs376516746 Chr.16: 21250337-21250337 on Build GRCh38
rs768421905 Chr.16: 21250414-21250414 on Build GRCh38
rs773311953 Chr.16: 21250450-21250450 on Build GRCh38
rs775043096 Chr.16: 21250310-21250310 on Build GRCh38
rs745438438 Chr.16: 21250299-21250299 on Build GRCh38
rs536102146 Chr.16: 21250314-21250314 on Build GRCh38
rs761017608 Chr.16: 21250399-21250399 on Build GRCh38
rs890943330 Chr.16: 21250271-21250276 on Build GRCh38
rs756469922 Chr.16: 21250270-21250270 on Build GRCh38
rs779384888 Chr.16: 21250297-21250297 on Build GRCh38

View Full Product Details