Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 147128596-147128867 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNTNAP2
Entrez Gene ID
26047
Gene Name
contactin associated protein-like 2
Gene Aliases
AUTS15, CASPR2, CDFE, NRXN4, PTHSL1
UniGene
Hs.655684
Gene Chromosome Location
Chr.7: 146116035-148420998 on Build GRCh38
Transcripts
NM_014141.5 XM_017011950.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
745047 c.949G>T p.G317* Chr.7: 147128702-147128702 on Build GRCh38
3879060 c.959C>A p.P320H Chr.7: 147128712-147128712 on Build GRCh38
745044 c.1032C>A p.G344G Chr.7: 147128785-147128785 on Build GRCh38
3635659 c.999C>T p.F333F Chr.7: 147128752-147128752 on Build GRCh38
3923036 c.1020C>T p.I340I Chr.7: 147128773-147128773 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs904267244 Chr.7: 147128791-147128791 on Build GRCh38
rs139042086 Chr.7: 147128718-147128718 on Build GRCh38
rs746335569 Chr.7: 147128781-147128781 on Build GRCh38
rs761393725 Chr.7: 147128747-147128748 on Build GRCh38
rs570190374 Chr.7: 147128831-147128831 on Build GRCh38
rs201927719 Chr.7: 147128656-147128656 on Build GRCh38
rs145832489 Chr.7: 147128786-147128786 on Build GRCh38
rs780045933 Chr.7: 147128786-147128793 on Build GRCh38
rs765211652 Chr.7: 147128716-147128716 on Build GRCh38
rs775112062 Chr.7: 147128664-147128664 on Build GRCh38
rs920771807 Chr.7: 147128634-147128634 on Build GRCh38
rs760724848 Chr.7: 147128715-147128715 on Build GRCh38
rs541651743 Chr.7: 147128649-147128649 on Build GRCh38
rs113086771 Chr.7: 147128711-147128711 on Build GRCh38
rs774172418 Chr.7: 147128680-147128680 on Build GRCh38
rs794727873 Chr.7: 147128832-147128832 on Build GRCh38
rs771980187 Chr.7: 147128818-147128818 on Build GRCh38
rs954835249 Chr.7: 147128719-147128719 on Build GRCh38
rs374812409 Chr.7: 147128676-147128676 on Build GRCh38
rs1016325711 Chr.7: 147128674-147128674 on Build GRCh38
rs139180845 Chr.7: 147128836-147128836 on Build GRCh38
rs760416207 Chr.7: 147128826-147128826 on Build GRCh38
rs768298723 Chr.7: 147128792-147128792 on Build GRCh38
rs1034170979 Chr.7: 147128815-147128815 on Build GRCh38
rs367878797 Chr.7: 147128783-147128783 on Build GRCh38
rs561532560 Chr.7: 147128653-147128653 on Build GRCh38
rs761646749 Chr.7: 147128696-147128696 on Build GRCh38
rs758429654 Chr.7: 147128736-147128736 on Build GRCh38
rs774427240 Chr.7: 147128797-147128797 on Build GRCh38
rs781608779 Chr.7: 147128655-147128655 on Build GRCh38
rs774084392 Chr.7: 147128798-147128798 on Build GRCh38
rs1041122943 Chr.7: 147128732-147128732 on Build GRCh38
rs930847514 Chr.7: 147128636-147128636 on Build GRCh38
rs765754117 Chr.7: 147128712-147128712 on Build GRCh38
rs773110260 Chr.7: 147128821-147128821 on Build GRCh38
rs369966545 Chr.7: 147128646-147128646 on Build GRCh38
rs750383045 Chr.7: 147128701-147128701 on Build GRCh38
rs201253086 Chr.7: 147128804-147128804 on Build GRCh38
rs781590630 Chr.7: 147128773-147128773 on Build GRCh38
rs146201964 Chr.7: 147128768-147128768 on Build GRCh38
rs115730262 Chr.7: 147128665-147128665 on Build GRCh38
rs794727872 Chr.7: 147128698-147128698 on Build GRCh38
rs748787013 Chr.7: 147128790-147128790 on Build GRCh38
rs769170569 Chr.7: 147128660-147128660 on Build GRCh38
rs764241663 Chr.7: 147128740-147128740 on Build GRCh38
rs142122012 Chr.7: 147128785-147128785 on Build GRCh38
rs767425362 Chr.7: 147128700-147128700 on Build GRCh38
rs757610472 Chr.7: 147128769-147128769 on Build GRCh38

View Full Product Details