Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 35766843-35767068 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PROSER3
Entrez Gene ID
148137
Gene Name
proline and serine rich 3
Gene Aliases
C19orf55
UniGene
Hs.527982
Gene Chromosome Location
Chr.19: 35758139-35770115 on Build GRCh38
Transcripts
NM_001039887.2 XM_011526528.1 XM_011526529.2 XM_011526530.1 XM_011526531.1 XM_011526532.1 XM_011526533.2 XM_011526534.1 XM_011526535.2 XM_011526536.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1392962 c.941C>A p.P314H Chr.19: 35766939-35766939 on Build GRCh38
6557786 c.923G>T p.W308L Chr.19: 35766921-35766921 on Build GRCh38
6557787 c.887G>T p.W296L Chr.19: 35766921-35766921 on Build GRCh38
6417169 c.900G>T p.Q300H Chr.19: 35766898-35766898 on Build GRCh38
6900423 c.911G>A p.G304D Chr.19: 35766909-35766909 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750121588 Chr.19: 35767006-35767006 on Build GRCh38
rs766278633 Chr.19: 35767038-35767040 on Build GRCh38
rs374887676 Chr.19: 35766875-35766875 on Build GRCh38
rs756957808 Chr.19: 35766891-35766891 on Build GRCh38
rs748701598 Chr.19: 35766927-35766927 on Build GRCh38
rs372740276 Chr.19: 35766884-35766884 on Build GRCh38
rs762947329 Chr.19: 35766966-35766966 on Build GRCh38
rs749494615 Chr.19: 35766928-35766928 on Build GRCh38
rs373793251 Chr.19: 35766988-35766988 on Build GRCh38
rs750440553 Chr.19: 35767032-35767035 on Build GRCh38
rs550616208 Chr.19: 35766897-35766897 on Build GRCh38
rs763997209 Chr.19: 35766969-35766969 on Build GRCh38
rs533506839 Chr.19: 35766947-35766947 on Build GRCh38
rs868567379 Chr.19: 35766995-35766995 on Build GRCh38
rs753991910 Chr.19: 35767034-35767035 on Build GRCh38
rs182004827 Chr.19: 35766962-35766962 on Build GRCh38
rs763667951 Chr.19: 35766879-35766879 on Build GRCh38
rs773891657 Chr.19: 35766972-35766972 on Build GRCh38
rs752230389 Chr.19: 35766919-35766919 on Build GRCh38
rs776874109 Chr.19: 35766951-35766951 on Build GRCh38
rs761448950 Chr.19: 35766977-35766977 on Build GRCh38
rs755425397 Chr.19: 35766913-35766913 on Build GRCh38
rs534195783 Chr.19: 35766976-35766976 on Build GRCh38
rs907280072 Chr.19: 35766922-35766922 on Build GRCh38
rs929440917 Chr.19: 35766923-35766923 on Build GRCh38
rs762316515 Chr.19: 35766878-35766878 on Build GRCh38
rs368917297 Chr.19: 35766876-35766876 on Build GRCh38
rs779238308 Chr.19: 35766924-35766924 on Build GRCh38
rs74172760 Chr.19: 35767033-35767034 on Build GRCh38
rs759372021 Chr.19: 35766870-35766870 on Build GRCh38
rs775226856 Chr.19: 35766963-35766963 on Build GRCh38
rs944705359 Chr.19: 35766983-35766983 on Build GRCh38
rs147473895 Chr.19: 35766982-35766982 on Build GRCh38
rs231219 Chr.19: 35766904-35766904 on Build GRCh38
rs57901537 Chr.19: 35767008-35767008 on Build GRCh38
rs1030536508 Chr.19: 35767026-35767026 on Build GRCh38
rs35876904 Chr.19: 35767032-35767033 on Build GRCh38
rs765637749 Chr.19: 35767028-35767028 on Build GRCh38
rs771115872 Chr.19: 35766950-35766950 on Build GRCh38
rs551664520 Chr.19: 35766948-35766948 on Build GRCh38

View Full Product Details