Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 98756555-98756794 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKS6
Entrez Gene ID
203286
Gene Name
ankyrin repeat and sterile alpha motif domain containing 6
Gene Aliases
ANKRD14, NPHP16, PKDR1, SAMD6
UniGene
Hs.406890
Gene Chromosome Location
Chr.9: 98732009-98796542 on Build GRCh38
Transcripts
NM_173551.4 XM_005251793.4 XM_005251794.4 XM_006716998.3 XM_006716999.2 XM_011518357.2 XM_017014445.1 XM_017014446.1 XM_017014447.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5012051 c.2177C>T p.T726I Chr.9: 98756569-98756569 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs937756780 Chr.9: 98756711-98756711 on Build GRCh38
rs763588317 Chr.9: 98756613-98756613 on Build GRCh38
rs577954143 Chr.9: 98756637-98756637 on Build GRCh38
rs146104372 Chr.9: 98756697-98756697 on Build GRCh38
rs184449242 Chr.9: 98756687-98756687 on Build GRCh38
rs896143096 Chr.9: 98756726-98756726 on Build GRCh38
rs372906336 Chr.9: 98756577-98756577 on Build GRCh38
rs201518041 Chr.9: 98756591-98756591 on Build GRCh38
rs376048873 Chr.9: 98756618-98756618 on Build GRCh38
rs752281992 Chr.9: 98756647-98756647 on Build GRCh38
rs923781913 Chr.9: 98756691-98756691 on Build GRCh38
rs752558829 Chr.9: 98756685-98756685 on Build GRCh38
rs371305410 Chr.9: 98756656-98756656 on Build GRCh38
rs1054824680 Chr.9: 98756721-98756721 on Build GRCh38
rs945317660 Chr.9: 98756672-98756672 on Build GRCh38
rs558159827 Chr.9: 98756635-98756635 on Build GRCh38
rs78858055 Chr.9: 98756640-98756640 on Build GRCh38

View Full Product Details