Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 45841741-45842012 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACOT8
Entrez Gene ID
10005
Gene Name
acyl-CoA thioesterase 8
Gene Aliases
HNAACTE, NAP1, PTE-1, PTE-2, PTE1, PTE2, hACTE-III, hTE
UniGene
Hs.444776
Gene Chromosome Location
Chr.20: 45841721-45857409 on Build GRCh38
Transcripts
NM_005469.3 XM_005260239.3 XM_011528480.2 NM_001042632.2 NM_001042633.2 NM_033421.3 NM_152897.2 XM_005260608.4 XM_011529095.2

Gene Symbol
SNX21
Entrez Gene ID
90203
Gene Name
sorting nexin family member 21
Gene Aliases
C20orf161, PP3993, SNX-L, dJ337O18.4
UniGene
Hs.472854
Gene Chromosome Location
Chr.20: 45833418-45843275 on Build GRCh38
Transcripts
NM_005469.3 XM_005260239.3 XM_011528480.2 NM_001042632.2 NM_001042633.2 NM_033421.3 NM_152897.2 XM_005260608.4 XM_011529095.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
318477 c.946G>A p.E316K Chr.20: 45841852-45841852 on Build GRCh38
6849927 c.916G>A p.G306S Chr.20: 45841882-45841882 on Build GRCh38
1641356 c.850C>T p.R284W Chr.20: 45841948-45841948 on Build GRCh38
4098872 c.869G>A p.R290Q Chr.20: 45841929-45841929 on Build GRCh38
1412087 c.842-2A>G p.? Chr.20: 45841958-45841958 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs938969921 Chr.20: 45841776-45841776 on Build GRCh38
rs150475004 Chr.20: 45841929-45841929 on Build GRCh38
rs778450892 Chr.20: 45841880-45841880 on Build GRCh38
rs775951683 Chr.20: 45841915-45841915 on Build GRCh38
rs147976812 Chr.20: 45841874-45841874 on Build GRCh38
rs927841572 Chr.20: 45841969-45841969 on Build GRCh38
rs145182943 Chr.20: 45841799-45841799 on Build GRCh38
rs778167110 Chr.20: 45841973-45841973 on Build GRCh38
rs73308468 Chr.20: 45841829-45841829 on Build GRCh38
rs758433775 Chr.20: 45841849-45841849 on Build GRCh38
rs374061878 Chr.20: 45841903-45841903 on Build GRCh38
rs768723547 Chr.20: 45841921-45841921 on Build GRCh38
rs1022152627 Chr.20: 45841937-45841937 on Build GRCh38
rs776641728 Chr.20: 45841922-45841922 on Build GRCh38
rs148674138 Chr.20: 45841936-45841936 on Build GRCh38
rs756482650 Chr.20: 45841961-45841961 on Build GRCh38
rs753668905 Chr.20: 45841825-45841825 on Build GRCh38
rs745573705 Chr.20: 45841986-45841986 on Build GRCh38
rs143518276 Chr.20: 45841873-45841873 on Build GRCh38
rs775135298 Chr.20: 45841915-45841915 on Build GRCh38
rs892504403 Chr.20: 45841854-45841854 on Build GRCh38
rs750615669 Chr.20: 45841836-45841836 on Build GRCh38
rs755473458 Chr.20: 45841871-45841871 on Build GRCh38
rs775337601 Chr.20: 45841818-45841818 on Build GRCh38
rs1052846247 Chr.20: 45841841-45841841 on Build GRCh38
rs771521337 Chr.20: 45841910-45841910 on Build GRCh38
rs113036472 Chr.20: 45841839-45841839 on Build GRCh38
rs764738668 Chr.20: 45841969-45841969 on Build GRCh38
rs978015529 Chr.20: 45841965-45841965 on Build GRCh38
rs761728365 Chr.20: 45841923-45841923 on Build GRCh38
rs771560534 Chr.20: 45841815-45841815 on Build GRCh38
rs963611521 Chr.20: 45841988-45841988 on Build GRCh38
rs367746396 Chr.20: 45841827-45841827 on Build GRCh38
rs763208454 Chr.20: 45841930-45841930 on Build GRCh38
rs760616849 Chr.20: 45841888-45841888 on Build GRCh38
rs760658642 Chr.20: 45841820-45841820 on Build GRCh38
rs745510468 Chr.20: 45841886-45841886 on Build GRCh38
rs775461604 Chr.20: 45841837-45841842 on Build GRCh38
rs752104246 Chr.20: 45841862-45841862 on Build GRCh38
rs766463181 Chr.20: 45841851-45841851 on Build GRCh38
rs368111029 Chr.20: 45841931-45841931 on Build GRCh38
rs764047505 Chr.20: 45841821-45841821 on Build GRCh38
rs994405666 Chr.20: 45841834-45841834 on Build GRCh38
rs147298395 Chr.20: 45841947-45841947 on Build GRCh38
rs753164804 Chr.20: 45841948-45841948 on Build GRCh38
rs138553906 Chr.20: 45841932-45841932 on Build GRCh38
rs1048913 Chr.20: 45841856-45841856 on Build GRCh38
rs941532972 Chr.20: 45841807-45841807 on Build GRCh38
rs770476823 Chr.20: 45841796-45841796 on Build GRCh38
rs760460112 Chr.20: 45841920-45841920 on Build GRCh38
rs754993050 Chr.20: 45841945-45841945 on Build GRCh38
rs141643903 Chr.20: 45841879-45841879 on Build GRCh38
rs536751648 Chr.20: 45841877-45841877 on Build GRCh38
rs774557310 Chr.20: 45841806-45841806 on Build GRCh38
rs763391361 Chr.20: 45841928-45841928 on Build GRCh38

View Full Product Details