Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 65157573-65157844 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLPX
Entrez Gene ID
10845
Gene Name
caseinolytic mitochondrial matrix peptidase chaperone subunit
Gene Aliases
-
UniGene
Hs.113823
Gene Chromosome Location
Chr.15: 65148219-65185420 on Build GRCh38
Transcripts
NM_006660.4 NR_133680.1 XM_011521164.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1180364 c.1003G>A p.V335M Chr.15: 65157800-65157800 on Build GRCh38
4564140 c.1020_1021CC>TT p.Q341* Chr.15: 65157782-65157783 on Build GRCh38
6630664 c.988G>A p.E330K Chr.15: 65157815-65157815 on Build GRCh38
4056270 c.1044A>C p.E348D Chr.15: 65157759-65157759 on Build GRCh38
5849549 c.1047delA p.A350fs*11 Chr.15: 65157756-65157756 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369618230 Chr.15: 65157695-65157695 on Build GRCh38
rs55657623 Chr.15: 65157696-65157696 on Build GRCh38
rs151195584 Chr.15: 65157771-65157771 on Build GRCh38
rs756514374 Chr.15: 65157808-65157808 on Build GRCh38
rs753829887 Chr.15: 65157753-65157753 on Build GRCh38
rs986809803 Chr.15: 65157675-65157675 on Build GRCh38
rs571223872 Chr.15: 65157722-65157722 on Build GRCh38
rs767700739 Chr.15: 65157731-65157731 on Build GRCh38
rs543425198 Chr.15: 65157658-65157658 on Build GRCh38
rs141791602 Chr.15: 65157679-65157679 on Build GRCh38
rs770263938 Chr.15: 65157705-65157705 on Build GRCh38
rs752543706 Chr.15: 65157743-65157743 on Build GRCh38
rs370130335 Chr.15: 65157725-65157725 on Build GRCh38
rs566819699 Chr.15: 65157676-65157676 on Build GRCh38
rs777188761 Chr.15: 65157624-65157624 on Build GRCh38
rs962216584 Chr.15: 65157644-65157644 on Build GRCh38
rs749923096 Chr.15: 65157768-65157768 on Build GRCh38
rs142142822 Chr.15: 65157765-65157765 on Build GRCh38
rs771395959 Chr.15: 65157819-65157819 on Build GRCh38
rs760511159 Chr.15: 65157745-65157745 on Build GRCh38
rs774373959 Chr.15: 65157723-65157723 on Build GRCh38
rs201645826 Chr.15: 65157769-65157769 on Build GRCh38
rs764875363 Chr.15: 65157766-65157766 on Build GRCh38
rs763308876 Chr.15: 65157715-65157715 on Build GRCh38
rs944935472 Chr.15: 65157604-65157604 on Build GRCh38
rs749120119 Chr.15: 65157646-65157646 on Build GRCh38
rs140343605 Chr.15: 65157809-65157809 on Build GRCh38
rs918687478 Chr.15: 65157663-65157665 on Build GRCh38
rs536717501 Chr.15: 65157748-65157748 on Build GRCh38
rs373159447 Chr.15: 65157777-65157777 on Build GRCh38
rs748528883 Chr.15: 65157792-65157792 on Build GRCh38
rs900808306 Chr.15: 65157671-65157671 on Build GRCh38
rs779841328 Chr.15: 65157775-65157775 on Build GRCh38
rs1016043750 Chr.15: 65157634-65157634 on Build GRCh38
rs773772968 Chr.15: 65157708-65157708 on Build GRCh38
rs111265035 Chr.15: 65157816-65157816 on Build GRCh38

View Full Product Details