Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 74575371-74575644 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
M1AP
Entrez Gene ID
130951
Gene Name
meiosis 1 associated protein
Gene Aliases
C2orf65, D6Mm5e, SPATA37
UniGene
Hs.348645
Gene Chromosome Location
Chr.2: 74557640-74648338 on Build GRCh38
Transcripts
NM_001281295.1 NM_001281296.1 NM_001321739.1 NM_138804.4 XM_005264152.3 XM_006711946.3 XM_011532548.2 XM_011532549.2 XM_011532550.2 XM_011532551.2 XM_011532552.2 XM_011532553.2 XM_011532554.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7277818 c.1034C>T p.T345I Chr.2: 74575478-74575478 on Build GRCh38
309710 c.1012C>A p.L338M Chr.2: 74575500-74575500 on Build GRCh38
1023079 c.944C>A p.S315Y Chr.2: 74575568-74575568 on Build GRCh38
1023078 c.949G>A p.G317R Chr.2: 74575563-74575563 on Build GRCh38
5805473 c.939A>T p.L313F Chr.2: 74575573-74575573 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762842420 Chr.2: 74575585-74575585 on Build GRCh38
rs140179344 Chr.2: 74575563-74575563 on Build GRCh38
rs1041226415 Chr.2: 74575591-74575591 on Build GRCh38
rs575443272 Chr.2: 74575555-74575555 on Build GRCh38
rs772900148 Chr.2: 74575449-74575449 on Build GRCh38
rs780362539 Chr.2: 74575407-74575407 on Build GRCh38
rs766552022 Chr.2: 74575608-74575608 on Build GRCh38
rs746573239 Chr.2: 74575561-74575561 on Build GRCh38
rs746681618 Chr.2: 74575428-74575439 on Build GRCh38
rs757231810 Chr.2: 74575523-74575523 on Build GRCh38
rs371314363 Chr.2: 74575560-74575560 on Build GRCh38
rs991155183 Chr.2: 74575417-74575417 on Build GRCh38
rs765098025 Chr.2: 74575574-74575574 on Build GRCh38
rs776177329 Chr.2: 74575540-74575541 on Build GRCh38
rs761300399 Chr.2: 74575583-74575583 on Build GRCh38
rs921666567 Chr.2: 74575396-74575396 on Build GRCh38
rs148385130 Chr.2: 74575531-74575531 on Build GRCh38
rs760374511 Chr.2: 74575452-74575452 on Build GRCh38
rs76586849 Chr.2: 74575401-74575401 on Build GRCh38
rs749723707 Chr.2: 74575408-74575408 on Build GRCh38
rs773609290 Chr.2: 74575618-74575619 on Build GRCh38
rs909032822 Chr.2: 74575427-74575427 on Build GRCh38
rs375136912 Chr.2: 74575432-74575432 on Build GRCh38
rs756317597 Chr.2: 74575535-74575535 on Build GRCh38
rs765017398 Chr.2: 74575503-74575503 on Build GRCh38
rs903319068 Chr.2: 74575620-74575620 on Build GRCh38
rs759211969 Chr.2: 74575568-74575568 on Build GRCh38
rs759389242 Chr.2: 74575491-74575491 on Build GRCh38
rs764083330 Chr.2: 74575589-74575589 on Build GRCh38
rs770323846 Chr.2: 74575597-74575600 on Build GRCh38
rs372111313 Chr.2: 74575410-74575410 on Build GRCh38
rs761190055 Chr.2: 74575430-74575430 on Build GRCh38
rs748666857 Chr.2: 74575554-74575554 on Build GRCh38
rs368219097 Chr.2: 74575402-74575402 on Build GRCh38
rs144517996 Chr.2: 74575549-74575549 on Build GRCh38
rs553419066 Chr.2: 74575421-74575421 on Build GRCh38
rs775194157 Chr.2: 74575580-74575580 on Build GRCh38
rs369280100 Chr.2: 74575459-74575459 on Build GRCh38
rs368627831 Chr.2: 74575538-74575538 on Build GRCh38
rs6546912 Chr.2: 74575601-74575601 on Build GRCh38
rs150724944 Chr.2: 74575532-74575532 on Build GRCh38
rs878956718 Chr.2: 74575542-74575542 on Build GRCh38
rs752708443 Chr.2: 74575509-74575509 on Build GRCh38
rs750447823 Chr.2: 74575599-74575599 on Build GRCh38
rs755405354 Chr.2: 74575545-74575545 on Build GRCh38
rs753559168 Chr.2: 74575483-74575483 on Build GRCh38
rs1036254820 Chr.2: 74575512-74575512 on Build GRCh38
rs749441207 Chr.2: 74575564-74575564 on Build GRCh38
rs907198461 Chr.2: 74575527-74575527 on Build GRCh38
rs940642349 Chr.2: 74575448-74575448 on Build GRCh38
rs768474471 Chr.2: 74575486-74575487 on Build GRCh38
rs56802236 Chr.2: 74575540-74575540 on Build GRCh38

View Full Product Details