Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 22415468-22415713 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTAGE1
Entrez Gene ID
64693
Gene Name
cutaneous T-cell lymphoma-associated antigen 1
Gene Aliases
CT21.1, CT21.2, CTAGE, CTAGE-1, CTAGE-2
UniGene
Hs.406709
Gene Chromosome Location
Chr.18: 22413601-22417915 on Build GRCh38
Transcripts
NM_172241.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2885089 c.2218C>T p.P740S Chr.18: 22415594-22415594 on Build GRCh38
3890796 c.2128C>T p.P710S Chr.18: 22415684-22415684 on Build GRCh38
6082297 c.2145T>A p.A715A Chr.18: 22415667-22415667 on Build GRCh38
7363123 c.2163T>A p.N721K Chr.18: 22415649-22415649 on Build GRCh38
2885097 c.2194C>A p.R732S Chr.18: 22415618-22415618 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745372676 Chr.18: 22415562-22415562 on Build GRCh38
rs369191131 Chr.18: 22415609-22415609 on Build GRCh38
rs755122856 Chr.18: 22415538-22415538 on Build GRCh38
rs752844586 Chr.18: 22415607-22415607 on Build GRCh38
rs775299041 Chr.18: 22415565-22415565 on Build GRCh38
rs772756882 Chr.18: 22415643-22415643 on Build GRCh38
rs761505835 Chr.18: 22415585-22415585 on Build GRCh38
rs761346791 Chr.18: 22415638-22415638 on Build GRCh38
rs774225595 Chr.18: 22415637-22415637 on Build GRCh38
rs749260943 Chr.18: 22415618-22415618 on Build GRCh38
rs561341105 Chr.18: 22415506-22415506 on Build GRCh38
rs754305080 Chr.18: 22415529-22415529 on Build GRCh38
rs753187904 Chr.18: 22415673-22415673 on Build GRCh38
rs771653179 Chr.18: 22415589-22415590 on Build GRCh38
rs868556407 Chr.18: 22415594-22415595 on Build GRCh38
rs772399855 Chr.18: 22415555-22415555 on Build GRCh38
rs769564729 Chr.18: 22415563-22415563 on Build GRCh38
rs779890809 Chr.18: 22415617-22415617 on Build GRCh38
rs564402673 Chr.18: 22415658-22415658 on Build GRCh38
rs754360939 Chr.18: 22415596-22415596 on Build GRCh38
rs375206227 Chr.18: 22415640-22415640 on Build GRCh38
rs768218761 Chr.18: 22415570-22415570 on Build GRCh38
rs756455321 Chr.18: 22415527-22415527 on Build GRCh38
rs372016465 Chr.18: 22415615-22415615 on Build GRCh38
rs766708550 Chr.18: 22415595-22415595 on Build GRCh38
rs1017290645 Chr.18: 22415619-22415619 on Build GRCh38
rs955983312 Chr.18: 22415679-22415679 on Build GRCh38
rs748449666 Chr.18: 22415546-22415546 on Build GRCh38
rs941517751 Chr.18: 22415507-22415507 on Build GRCh38
rs778001588 Chr.18: 22415556-22415556 on Build GRCh38
rs773605411 Chr.18: 22415654-22415654 on Build GRCh38
rs1045555239 Chr.18: 22415532-22415532 on Build GRCh38
rs758421486 Chr.18: 22415675-22415675 on Build GRCh38
rs773818348 Chr.18: 22415584-22415584 on Build GRCh38
rs762344027 Chr.18: 22415595-22415596 on Build GRCh38
rs1044488727 Chr.18: 22415504-22415504 on Build GRCh38
rs745680070 Chr.18: 22415557-22415557 on Build GRCh38
rs369655419 Chr.18: 22415666-22415666 on Build GRCh38
rs751791912 Chr.18: 22415682-22415682 on Build GRCh38
rs550845102 Chr.18: 22415587-22415587 on Build GRCh38
rs770098578 Chr.18: 22415596-22415596 on Build GRCh38
rs779124305 Chr.18: 22415541-22415541 on Build GRCh38
rs779175519 Chr.18: 22415601-22415601 on Build GRCh38
rs760363760 Chr.18: 22415648-22415648 on Build GRCh38
rs948790194 Chr.18: 22415496-22415496 on Build GRCh38
rs376613924 Chr.18: 22415676-22415676 on Build GRCh38
rs780459760 Chr.18: 22415528-22415528 on Build GRCh38
rs755538577 Chr.18: 22415597-22415597 on Build GRCh38
rs375036881 Chr.18: 22415569-22415569 on Build GRCh38
rs373446707 Chr.18: 22415671-22415671 on Build GRCh38
rs777304069 Chr.18: 22415590-22415590 on Build GRCh38
rs768556331 Chr.18: 22415635-22415635 on Build GRCh38

View Full Product Details