Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 215234753-215234988 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNK2
Entrez Gene ID
3776
Gene Name
potassium two pore domain channel subfamily K member 2
Gene Aliases
K2p2.1, TPKC1, TREK, TREK-1, TREK1, hTREK-1c, hTREK-1e
UniGene
Hs.497745
Gene Chromosome Location
Chr.1: 215002920-215237093 on Build GRCh38
Transcripts
NM_001017424.2 NM_001017425.2 NM_014217.3 XM_011509522.2 XM_011509523.1 XM_011509524.2 XM_017001248.1 XM_017001249.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
903793 c.1063G>A p.A355T Chr.1: 215234972-215234972 on Build GRCh38
6124469 c.1074C>T p.A358A Chr.1: 215234938-215234938 on Build GRCh38
6541713 c.1056G>T p.Q352H Chr.1: 215234932-215234932 on Build GRCh38
4405040 c.959A>C p.N320T Chr.1: 215234868-215234868 on Build GRCh38
1211567 c.1065C>A p.F355L Chr.1: 215234929-215234929 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769208249 Chr.1: 215234961-215234961 on Build GRCh38
rs897321993 Chr.1: 215234849-215234849 on Build GRCh38
rs371990293 Chr.1: 215234901-215234901 on Build GRCh38
rs777330377 Chr.1: 215234917-215234917 on Build GRCh38
rs751828549 Chr.1: 215234878-215234878 on Build GRCh38
rs753784478 Chr.1: 215234876-215234876 on Build GRCh38
rs774425336 Chr.1: 215234819-215234819 on Build GRCh38
rs548027569 Chr.1: 215234899-215234899 on Build GRCh38
rs755353413 Chr.1: 215234817-215234818 on Build GRCh38
rs766296960 Chr.1: 215234869-215234869 on Build GRCh38
rs768684879 Chr.1: 215234805-215234805 on Build GRCh38
rs764785599 Chr.1: 215234886-215234886 on Build GRCh38
rs376567893 Chr.1: 215234792-215234792 on Build GRCh38
rs770303631 Chr.1: 215234790-215234790 on Build GRCh38
rs566476827 Chr.1: 215234957-215234957 on Build GRCh38
rs773024551 Chr.1: 215234848-215234848 on Build GRCh38
rs772248987 Chr.1: 215234844-215234844 on Build GRCh38
rs376273447 Chr.1: 215234825-215234825 on Build GRCh38
rs369548068 Chr.1: 215234879-215234879 on Build GRCh38
rs778959218 Chr.1: 215234962-215234962 on Build GRCh38
rs780429822 Chr.1: 215234956-215234956 on Build GRCh38
rs749817683 Chr.1: 215234794-215234794 on Build GRCh38
rs760432472 Chr.1: 215234857-215234857 on Build GRCh38
rs750778633 Chr.1: 215234929-215234929 on Build GRCh38
rs746288660 Chr.1: 215234789-215234789 on Build GRCh38
rs762080983 Chr.1: 215234828-215234828 on Build GRCh38
rs781610476 Chr.1: 215234787-215234787 on Build GRCh38
rs113654678 Chr.1: 215234863-215234863 on Build GRCh38

View Full Product Details